Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49808193-49808458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUZ
Entrez Gene ID
80199
Gene Name
fuzzy planar cell polarity protein
Gene Aliases
FY, NTD
UniGene
Hs.288800
Gene Chromosome Location
Chr.19: 49806866-49817376 on Build GRCh38
Transcripts
NM_001171937.1 NM_025129.4 NR_033269.1 XM_006723399.3 XM_011527339.1 XM_011527340.1 XM_011527341.2 XM_011527342.1 XM_011527343.1 XM_011527345.1 XM_011527346.1 XM_011527347.1 XM_017027319.1 XM_017027320.1 XM_017027321.1 XM_017027322.1 XM_017027323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3536970 c.991C>T p.R331* Chr.19: 49808456-49808456 on Build GRCh38
4080200 c.1020G>A p.T340T Chr.19: 49808427-49808427 on Build GRCh38
6056744 c.1013C>T p.T338I Chr.19: 49808434-49808434 on Build GRCh38
6667949 c.992G>A p.R331Q Chr.19: 49808455-49808455 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030873185 Chr.19: 49808311-49808311 on Build GRCh38
rs781077490 Chr.19: 49808388-49808388 on Build GRCh38
rs983468722 Chr.19: 49808355-49808355 on Build GRCh38
rs555228691 Chr.19: 49808365-49808365 on Build GRCh38
rs138765988 Chr.19: 49808407-49808407 on Build GRCh38
rs368396623 Chr.19: 49808397-49808397 on Build GRCh38
rs186368010 Chr.19: 49808279-49808279 on Build GRCh38
rs909232091 Chr.19: 49808361-49808361 on Build GRCh38
rs889617903 Chr.19: 49808268-49808268 on Build GRCh38
rs569587505 Chr.19: 49808294-49808294 on Build GRCh38
rs756079112 Chr.19: 49808393-49808393 on Build GRCh38
rs998875511 Chr.19: 49808362-49808362 on Build GRCh38
rs771338095 Chr.19: 49808416-49808416 on Build GRCh38
rs534754038 Chr.19: 49808411-49808411 on Build GRCh38
rs1006823480 Chr.19: 49808226-49808226 on Build GRCh38
rs777665466 Chr.19: 49808263-49808263 on Build GRCh38
rs758150221 Chr.19: 49808370-49808371 on Build GRCh38
rs995372976 Chr.19: 49808231-49808232 on Build GRCh38
rs770532530 Chr.19: 49808408-49808408 on Build GRCh38
rs748265542 Chr.19: 49808321-49808321 on Build GRCh38
rs751862735 Chr.19: 49808378-49808378 on Build GRCh38
rs745576184 Chr.19: 49808426-49808426 on Build GRCh38
rs376806325 Chr.19: 49808389-49808389 on Build GRCh38
rs1018165245 Chr.19: 49808270-49808270 on Build GRCh38
rs766520008 Chr.19: 49808376-49808376 on Build GRCh38
rs777975622 Chr.19: 49808395-49808395 on Build GRCh38
rs754722600 Chr.19: 49808383-49808383 on Build GRCh38
rs528642796 Chr.19: 49808320-49808320 on Build GRCh38
rs1028138496 Chr.19: 49808254-49808254 on Build GRCh38

View Full Product Details