Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19283276-19283539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN2
Entrez Gene ID
22905
Gene Name
epsin 2
Gene Aliases
EHB21
UniGene
Hs.743983
Gene Chromosome Location
Chr.17: 19237377-19336715 on Build GRCh38
Transcripts
NM_001102664.1 NM_014964.4 NM_148921.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1679640 c.212G>A p.W71* Chr.17: 19283331-19283331 on Build GRCh38
1195499 c.268G>A p.E90K Chr.17: 19283387-19283387 on Build GRCh38
4681407 c.371G>A p.R124H Chr.17: 19283490-19283490 on Build GRCh38
3515022 c.412G>A p.E138K Chr.17: 19283531-19283531 on Build GRCh38
6145942 c.206A>C p.K69T Chr.17: 19283325-19283325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55641774 Chr.17: 19283445-19283445 on Build GRCh38
rs772783424 Chr.17: 19283478-19283478 on Build GRCh38
rs369710969 Chr.17: 19283383-19283383 on Build GRCh38
rs747758321 Chr.17: 19283473-19283473 on Build GRCh38
rs774000263 Chr.17: 19283460-19283460 on Build GRCh38
rs778205804 Chr.17: 19283298-19283298 on Build GRCh38
rs949748262 Chr.17: 19283372-19283372 on Build GRCh38
rs1042000504 Chr.17: 19283433-19283433 on Build GRCh38
rs776575965 Chr.17: 19283486-19283486 on Build GRCh38
rs371807837 Chr.17: 19283425-19283425 on Build GRCh38
rs752850441 Chr.17: 19283417-19283417 on Build GRCh38
rs760440547 Chr.17: 19283400-19283400 on Build GRCh38
rs369019354 Chr.17: 19283426-19283426 on Build GRCh38
rs200979660 Chr.17: 19283307-19283307 on Build GRCh38
rs777052359 Chr.17: 19283338-19283338 on Build GRCh38
rs753787529 Chr.17: 19283409-19283409 on Build GRCh38
rs777951481 Chr.17: 19283423-19283423 on Build GRCh38
rs768221067 Chr.17: 19283453-19283453 on Build GRCh38
rs767350047 Chr.17: 19283390-19283390 on Build GRCh38
rs764344854 Chr.17: 19283516-19283516 on Build GRCh38
rs767335671 Chr.17: 19283397-19283397 on Build GRCh38
rs116241641 Chr.17: 19283497-19283497 on Build GRCh38
rs139687943 Chr.17: 19283489-19283489 on Build GRCh38
rs768345994 Chr.17: 19283367-19283367 on Build GRCh38
rs184266158 Chr.17: 19283515-19283515 on Build GRCh38
rs766147588 Chr.17: 19283484-19283484 on Build GRCh38
rs747434950 Chr.17: 19283308-19283308 on Build GRCh38
rs544825853 Chr.17: 19283434-19283434 on Build GRCh38
rs778979636 Chr.17: 19283415-19283415 on Build GRCh38
rs142873641 Chr.17: 19283350-19283350 on Build GRCh38
rs750259068 Chr.17: 19283391-19283391 on Build GRCh38
rs147412182 Chr.17: 19283446-19283446 on Build GRCh38
rs199887915 Chr.17: 19283386-19283386 on Build GRCh38
rs903112841 Chr.17: 19283513-19283513 on Build GRCh38
rs771491050 Chr.17: 19283320-19283320 on Build GRCh38
rs531286035 Chr.17: 19283408-19283408 on Build GRCh38
rs564542502 Chr.17: 19283482-19283482 on Build GRCh38
rs758650807 Chr.17: 19283422-19283422 on Build GRCh38

View Full Product Details