Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 64614895-64615136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7B1
Entrez Gene ID
9420
Gene Name
cytochrome P450 family 7 subfamily B member 1
Gene Aliases
CBAS3, CP7B, SPG5A
UniGene
Hs.657330
Gene Chromosome Location
Chr.8: 64590851-64798791 on Build GRCh38
Transcripts
NM_001324112.1 NM_004820.4 XM_017014002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268121 c.960C>T p.D320D Chr.8: 64615123-64615123 on Build GRCh38
6273918 c.983C>A p.S328* Chr.8: 64615100-64615100 on Build GRCh38
6268061 c.955C>A p.R319S Chr.8: 64615128-64615128 on Build GRCh38
7259508 c.987A>G p.T329T Chr.8: 64615096-64615096 on Build GRCh38
4676046 c.1057G>A p.E353K Chr.8: 64615026-64615026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766523749 Chr.8: 64615026-64615026 on Build GRCh38
rs1004981575 Chr.8: 64615088-64615088 on Build GRCh38
rs370051597 Chr.8: 64614985-64614985 on Build GRCh38
rs890613012 Chr.8: 64615067-64615067 on Build GRCh38
rs752857874 Chr.8: 64615038-64615038 on Build GRCh38
rs759809468 Chr.8: 64615035-64615035 on Build GRCh38
rs59035258 Chr.8: 64615112-64615112 on Build GRCh38
rs761630724 Chr.8: 64615000-64615000 on Build GRCh38
rs757698965 Chr.8: 64615065-64615065 on Build GRCh38
rs771797585 Chr.8: 64615076-64615076 on Build GRCh38
rs773969365 Chr.8: 64614935-64614935 on Build GRCh38
rs768687788 Chr.8: 64614987-64614987 on Build GRCh38
rs1051991067 Chr.8: 64614963-64614963 on Build GRCh38
rs766682920 Chr.8: 64614942-64614943 on Build GRCh38
rs779409818 Chr.8: 64615077-64615077 on Build GRCh38
rs773055558 Chr.8: 64615113-64615113 on Build GRCh38
rs917696140 Chr.8: 64615105-64615105 on Build GRCh38
rs779479164 Chr.8: 64615066-64615066 on Build GRCh38
rs913277056 Chr.8: 64614955-64614955 on Build GRCh38
rs919597624 Chr.8: 64614974-64614974 on Build GRCh38
rs1036508707 Chr.8: 64615103-64615103 on Build GRCh38
rs751526012 Chr.8: 64615004-64615005 on Build GRCh38
rs746555297 Chr.8: 64615084-64615084 on Build GRCh38
rs535728519 Chr.8: 64615047-64615047 on Build GRCh38
rs1057518485 Chr.8: 64615052-64615052 on Build GRCh38
rs568659813 Chr.8: 64614961-64614961 on Build GRCh38
rs964003792 Chr.8: 64614932-64614932 on Build GRCh38
rs767487623 Chr.8: 64615037-64615037 on Build GRCh38
rs745511463 Chr.8: 64615069-64615069 on Build GRCh38
rs145292008 Chr.8: 64615039-64615039 on Build GRCh38
rs776243938 Chr.8: 64614997-64614997 on Build GRCh38
rs778365454 Chr.8: 64615046-64615046 on Build GRCh38
rs768442659 Chr.8: 64615096-64615096 on Build GRCh38
rs947511932 Chr.8: 64614956-64614956 on Build GRCh38
rs751791001 Chr.8: 64615034-64615034 on Build GRCh38
rs1043417191 Chr.8: 64614960-64614960 on Build GRCh38
rs61745661 Chr.8: 64615051-64615051 on Build GRCh38
rs932223462 Chr.8: 64614962-64614962 on Build GRCh38
rs988881677 Chr.8: 64614950-64614950 on Build GRCh38

View Full Product Details