Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35459242-35459501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCE
Entrez Gene ID
2178
Gene Name
Fanconi anemia complementation group E
Gene Aliases
FACE, FAE
UniGene
Hs.302003
Gene Chromosome Location
Chr.6: 35452339-35467106 on Build GRCh38
Transcripts
NM_021922.2 XM_005248885.2 XM_005248886.2 XM_005248887.2 XM_005248888.3 XM_011514343.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626005 c.1189C>T p.P397S Chr.6: 35459406-35459406 on Build GRCh38
1078355 c.1159C>A p.L387M Chr.6: 35459376-35459376 on Build GRCh38
1730760 c.1114A>G p.I372V Chr.6: 35459331-35459331 on Build GRCh38
6447355 c.1140C>A p.S380S Chr.6: 35459357-35459357 on Build GRCh38
4992718 c.1125G>A p.L375L Chr.6: 35459342-35459342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534738858 Chr.6: 35459470-35459470 on Build GRCh38
rs1045372290 Chr.6: 35459305-35459305 on Build GRCh38
rs774510318 Chr.6: 35459314-35459314 on Build GRCh38
rs780820502 Chr.6: 35459350-35459350 on Build GRCh38
rs201500086 Chr.6: 35459432-35459432 on Build GRCh38
rs112375664 Chr.6: 35459271-35459271 on Build GRCh38
rs747040808 Chr.6: 35459475-35459476 on Build GRCh38
rs752130850 Chr.6: 35459438-35459438 on Build GRCh38
rs758622556 Chr.6: 35459380-35459382 on Build GRCh38
rs755688409 Chr.6: 35459474-35459474 on Build GRCh38
rs757769907 Chr.6: 35459454-35459454 on Build GRCh38
rs748733012 Chr.6: 35459479-35459479 on Build GRCh38
rs771924901 Chr.6: 35459398-35459398 on Build GRCh38
rs758792926 Chr.6: 35459303-35459303 on Build GRCh38
rs536294412 Chr.6: 35459292-35459292 on Build GRCh38
rs576468163 Chr.6: 35459372-35459372 on Build GRCh38
rs750812971 Chr.6: 35459473-35459473 on Build GRCh38
rs759843375 Chr.6: 35459290-35459290 on Build GRCh38
rs768455098 Chr.6: 35459375-35459375 on Build GRCh38
rs371020401 Chr.6: 35459358-35459358 on Build GRCh38
rs773156225 Chr.6: 35459399-35459399 on Build GRCh38
rs763145289 Chr.6: 35459418-35459418 on Build GRCh38
rs368422520 Chr.6: 35459327-35459327 on Build GRCh38
rs778387990 Chr.6: 35459308-35459308 on Build GRCh38
rs774487639 Chr.6: 35459378-35459378 on Build GRCh38
rs890942546 Chr.6: 35459423-35459423 on Build GRCh38
rs759782690 Chr.6: 35459406-35459406 on Build GRCh38
rs781483942 Chr.6: 35459472-35459472 on Build GRCh38
rs143234424 Chr.6: 35459333-35459333 on Build GRCh38
rs374232967 Chr.6: 35459359-35459359 on Build GRCh38
rs878854342 Chr.6: 35459323-35459323 on Build GRCh38
rs370897800 Chr.6: 35459367-35459367 on Build GRCh38
rs113175592 Chr.6: 35459337-35459337 on Build GRCh38
rs950005468 Chr.6: 35459280-35459280 on Build GRCh38
rs745461880 Chr.6: 35459356-35459356 on Build GRCh38
rs759721509 Chr.6: 35459420-35459420 on Build GRCh38
rs375385250 Chr.6: 35459281-35459281 on Build GRCh38
rs1038501708 Chr.6: 35459339-35459341 on Build GRCh38
rs764498332 Chr.6: 35459431-35459431 on Build GRCh38
rs375477547 Chr.6: 35459373-35459373 on Build GRCh38
rs752689765 Chr.6: 35459417-35459417 on Build GRCh38
rs765529209 Chr.6: 35459412-35459412 on Build GRCh38
rs778204394 Chr.6: 35459384-35459386 on Build GRCh38
rs761693769 Chr.6: 35459386-35459386 on Build GRCh38
rs752845362 Chr.6: 35459295-35459295 on Build GRCh38
rs34304743 Chr.6: 35459404-35459405 on Build GRCh38

View Full Product Details