Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158127236-158127493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM198B
Entrez Gene ID
51313
Gene Name
family with sequence similarity 198 member B
Gene Aliases
AD021, AD036, C4orf18
UniGene
Hs.567498
Gene Chromosome Location
Chr.4: 158124580-158173050 on Build GRCh38
Transcripts
NM_001031700.2 NM_001128424.1 NM_016613.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4569074 c.1500T>C p.L500L Chr.4: 158127491-158127491 on Build GRCh38
6406029 c.1579G>T p.E527* Chr.4: 158127412-158127412 on Build GRCh38
1594383 c.1554C>T p.H518H Chr.4: 158127437-158127437 on Build GRCh38
1428271 c.1531C>A p.L511I Chr.4: 158127460-158127460 on Build GRCh38
207301 c.1479C>T p.I493I Chr.4: 158127488-158127488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896365408 Chr.4: 158127434-158127434 on Build GRCh38
rs766627773 Chr.4: 158127302-158127302 on Build GRCh38
rs761669063 Chr.4: 158127442-158127442 on Build GRCh38
rs138391669 Chr.4: 158127451-158127451 on Build GRCh38
rs554024335 Chr.4: 158127386-158127386 on Build GRCh38
rs765326669 Chr.4: 158127442-158127442 on Build GRCh38
rs753444078 Chr.4: 158127359-158127359 on Build GRCh38
rs373417820 Chr.4: 158127377-158127377 on Build GRCh38
rs546145731 Chr.4: 158127401-158127401 on Build GRCh38
rs756991646 Chr.4: 158127365-158127365 on Build GRCh38
rs537536723 Chr.4: 158127323-158127323 on Build GRCh38
rs200022714 Chr.4: 158127436-158127436 on Build GRCh38
rs770046119 Chr.4: 158127413-158127413 on Build GRCh38
rs992411307 Chr.4: 158127317-158127317 on Build GRCh38
rs768918940 Chr.4: 158127399-158127399 on Build GRCh38
rs554697817 Chr.4: 158127341-158127341 on Build GRCh38
rs150200552 Chr.4: 158127268-158127268 on Build GRCh38
rs139005471 Chr.4: 158127452-158127452 on Build GRCh38
rs577182950 Chr.4: 158127392-158127392 on Build GRCh38
rs770954570 Chr.4: 158127423-158127423 on Build GRCh38
rs778703170 Chr.4: 158127373-158127373 on Build GRCh38
rs760864338 Chr.4: 158127293-158127293 on Build GRCh38
rs371939105 Chr.4: 158127385-158127385 on Build GRCh38
rs775290972 Chr.4: 158127426-158127426 on Build GRCh38
rs144840777 Chr.4: 158127444-158127444 on Build GRCh38
rs932224635 Chr.4: 158127296-158127296 on Build GRCh38
rs149442120 Chr.4: 158127437-158127437 on Build GRCh38
rs780739145 Chr.4: 158127391-158127391 on Build GRCh38
rs562853893 Chr.4: 158127420-158127420 on Build GRCh38
rs914591335 Chr.4: 158127306-158127306 on Build GRCh38
rs750057950 Chr.4: 158127450-158127450 on Build GRCh38
rs138943179 Chr.4: 158127375-158127375 on Build GRCh38
rs113800154 Chr.4: 158127417-158127417 on Build GRCh38
rs574453172 Chr.4: 158127349-158127349 on Build GRCh38
rs144055108 Chr.4: 158127433-158127433 on Build GRCh38
rs748461092 Chr.4: 158127410-158127410 on Build GRCh38

View Full Product Details