Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 256000-256229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20C
Entrez Gene ID
56975
Gene Name
family with sequence similarity 20 member C
Gene Aliases
DMP-4, DMP4, GEF-CK, RNS
UniGene
Hs.134742
Gene Chromosome Location
Chr.7: 192939-260774 on Build GRCh38
Transcripts
NM_020223.3 XM_017012450.1 XM_017012451.1 XM_017012455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6241133 c.1239G>T p.K413N Chr.7: 256015-256015 on Build GRCh38
1206306 c.1225C>T p.R409C Chr.7: 256001-256001 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187972052 Chr.7: 256049-256049 on Build GRCh38
rs929467036 Chr.7: 256153-256153 on Build GRCh38
rs932393277 Chr.7: 256037-256037 on Build GRCh38
rs552178274 Chr.7: 256116-256116 on Build GRCh38
rs1006617471 Chr.7: 256054-256054 on Build GRCh38
rs775008063 Chr.7: 256070-256070 on Build GRCh38
rs575909221 Chr.7: 256175-256175 on Build GRCh38
rs747591791 Chr.7: 256110-256110 on Build GRCh38
rs976431611 Chr.7: 256117-256117 on Build GRCh38
rs771584191 Chr.7: 256176-256176 on Build GRCh38
rs983622744 Chr.7: 256111-256111 on Build GRCh38
rs376163402 Chr.7: 256049-256049 on Build GRCh38
rs769194435 Chr.7: 256059-256059 on Build GRCh38
rs569922710 Chr.7: 256038-256038 on Build GRCh38
rs537319147 Chr.7: 256108-256108 on Build GRCh38
rs758159748 Chr.7: 256017-256017 on Build GRCh38
rs370300921 Chr.7: 256058-256058 on Build GRCh38
rs544981578 Chr.7: 256182-256182 on Build GRCh38
rs191110079 Chr.7: 256090-256090 on Build GRCh38
rs957749180 Chr.7: 256097-256097 on Build GRCh38
rs1023397465 Chr.7: 256078-256078 on Build GRCh38
rs140048285 Chr.7: 256093-256093 on Build GRCh38
rs146265893 Chr.7: 256048-256048 on Build GRCh38
rs374732415 Chr.7: 256073-256073 on Build GRCh38
rs963972147 Chr.7: 256133-256133 on Build GRCh38
rs923448848 Chr.7: 256151-256151 on Build GRCh38
rs923417798 Chr.7: 256023-256023 on Build GRCh38
rs999478738 Chr.7: 256092-256092 on Build GRCh38
rs986535444 Chr.7: 256191-256191 on Build GRCh38
rs774907032 Chr.7: 256064-256064 on Build GRCh38
rs895110902 Chr.7: 256204-256204 on Build GRCh38
rs574332586 Chr.7: 256055-256055 on Build GRCh38
rs74203528 Chr.7: 256185-256185 on Build GRCh38
rs1055553538 Chr.7: 256190-256190 on Build GRCh38
rs1024818745 Chr.7: 256069-256069 on Build GRCh38

View Full Product Details