Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21918700-21918959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC1
Entrez Gene ID
64215
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Gene Aliases
DNAJL1, ERdj1, HTJ1, MTJ1
UniGene
Hs.499000
Gene Chromosome Location
Chr.10: 21756548-22003768 on Build GRCh38
Transcripts
NM_022365.3 XM_011519614.2 XM_017016536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1730956 c.729+1G>T p.? Chr.10: 21918778-21918778 on Build GRCh38
6647161 c.698T>C p.L233P Chr.10: 21918810-21918810 on Build GRCh38
6065503 c.661G>C p.D221H Chr.10: 21918847-21918847 on Build GRCh38
2133079 c.658C>T p.H220Y Chr.10: 21918850-21918850 on Build GRCh38
4938324 c.657G>A p.W219* Chr.10: 21918851-21918851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868457628 Chr.10: 21918807-21918807 on Build GRCh38
rs989525665 Chr.10: 21918824-21918824 on Build GRCh38
rs777990309 Chr.10: 21918747-21918747 on Build GRCh38
rs765786136 Chr.10: 21918731-21918731 on Build GRCh38
rs761629189 Chr.10: 21918829-21918829 on Build GRCh38
rs768581529 Chr.10: 21918823-21918823 on Build GRCh38
rs773879751 Chr.10: 21918733-21918733 on Build GRCh38
rs763380206 Chr.10: 21918870-21918870 on Build GRCh38
rs943431959 Chr.10: 21918742-21918742 on Build GRCh38
rs771735296 Chr.10: 21918835-21918835 on Build GRCh38
rs375432713 Chr.10: 21918844-21918844 on Build GRCh38
rs916325217 Chr.10: 21918801-21918801 on Build GRCh38
rs749176575 Chr.10: 21918818-21918818 on Build GRCh38
rs370655862 Chr.10: 21918836-21918836 on Build GRCh38
rs758866421 Chr.10: 21918746-21918746 on Build GRCh38
rs140168773 Chr.10: 21918863-21918863 on Build GRCh38
rs367957606 Chr.10: 21918845-21918845 on Build GRCh38
rs939013326 Chr.10: 21918770-21918770 on Build GRCh38
rs1055238685 Chr.10: 21918788-21918788 on Build GRCh38
rs780266933 Chr.10: 21918911-21918911 on Build GRCh38
rs751891454 Chr.10: 21918758-21918758 on Build GRCh38
rs765874373 Chr.10: 21918855-21918855 on Build GRCh38
rs749323418 Chr.10: 21918913-21918913 on Build GRCh38
rs368727529 Chr.10: 21918917-21918917 on Build GRCh38
rs745965765 Chr.10: 21918785-21918785 on Build GRCh38
rs939613428 Chr.10: 21918898-21918898 on Build GRCh38
rs372656858 Chr.10: 21918918-21918918 on Build GRCh38
rs190541788 Chr.10: 21918837-21918837 on Build GRCh38
rs753175372 Chr.10: 21918740-21918740 on Build GRCh38
rs377284539 Chr.10: 21918812-21918812 on Build GRCh38
rs760220559 Chr.10: 21918849-21918849 on Build GRCh38
rs781520063 Chr.10: 21918772-21918772 on Build GRCh38
rs370406148 Chr.10: 21918901-21918901 on Build GRCh38
rs374226687 Chr.10: 21918910-21918910 on Build GRCh38
rs562480281 Chr.10: 21918769-21918769 on Build GRCh38
rs774023774 Chr.10: 21918825-21918825 on Build GRCh38
rs751898546 Chr.10: 21918888-21918888 on Build GRCh38
rs764591739 Chr.10: 21918872-21918872 on Build GRCh38
rs769971818 Chr.10: 21918800-21918800 on Build GRCh38
rs915364427 Chr.10: 21918830-21918830 on Build GRCh38

View Full Product Details