Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96266993-96267266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK3
Entrez Gene ID
2004
Gene Name
ELK3, ETS transcription factor
Gene Aliases
ERP, NET, SAP-2, SAP2
UniGene
Hs.46523
Gene Chromosome Location
Chr.12: 96194382-96269835 on Build GRCh38
Transcripts
NM_001303511.1 NM_005230.3 XM_006719275.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5796509 c.1164C>A p.I388I Chr.12: 96267120-96267120 on Build GRCh38
944737 c.1215G>T p.Q405H Chr.12: 96267171-96267171 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138930220 Chr.12: 96267146-96267146 on Build GRCh38
rs760335647 Chr.12: 96267193-96267193 on Build GRCh38
rs766886724 Chr.12: 96267151-96267153 on Build GRCh38
rs368507409 Chr.12: 96267152-96267152 on Build GRCh38
rs142164746 Chr.12: 96267148-96267148 on Build GRCh38
rs374036562 Chr.12: 96267185-96267185 on Build GRCh38
rs1044612532 Chr.12: 96267024-96267025 on Build GRCh38
rs891531766 Chr.12: 96267233-96267233 on Build GRCh38
rs75958097 Chr.12: 96267207-96267207 on Build GRCh38
rs200436208 Chr.12: 96267076-96267076 on Build GRCh38
rs762959909 Chr.12: 96267172-96267172 on Build GRCh38
rs766199561 Chr.12: 96267095-96267095 on Build GRCh38
rs771351901 Chr.12: 96267050-96267050 on Build GRCh38
rs754403067 Chr.12: 96267122-96267122 on Build GRCh38
rs752368172 Chr.12: 96267114-96267114 on Build GRCh38
rs778466030 Chr.12: 96267036-96267036 on Build GRCh38
rs201670464 Chr.12: 96267184-96267184 on Build GRCh38
rs964848817 Chr.12: 96267227-96267227 on Build GRCh38
rs1012054572 Chr.12: 96267124-96267124 on Build GRCh38
rs142850366 Chr.12: 96267105-96267105 on Build GRCh38
rs751253932 Chr.12: 96267096-96267096 on Build GRCh38
rs759391172 Chr.12: 96267099-96267099 on Build GRCh38
rs776505682 Chr.12: 96267163-96267163 on Build GRCh38
rs562893955 Chr.12: 96267037-96267037 on Build GRCh38
rs772390876 Chr.12: 96267144-96267144 on Build GRCh38
rs145057515 Chr.12: 96267129-96267129 on Build GRCh38
rs151047138 Chr.12: 96267206-96267206 on Build GRCh38
rs760780471 Chr.12: 96267088-96267088 on Build GRCh38
rs908570883 Chr.12: 96267181-96267181 on Build GRCh38
rs904309149 Chr.12: 96267025-96267025 on Build GRCh38
rs367548711 Chr.12: 96267187-96267187 on Build GRCh38
rs758779228 Chr.12: 96267225-96267225 on Build GRCh38
rs2302901 Chr.12: 96267236-96267236 on Build GRCh38
rs541968932 Chr.12: 96267118-96267118 on Build GRCh38
rs777371931 Chr.12: 96267121-96267121 on Build GRCh38
rs763688520 Chr.12: 96267197-96267197 on Build GRCh38
rs371199581 Chr.12: 96267138-96267138 on Build GRCh38
rs774542286 Chr.12: 96267051-96267051 on Build GRCh38
rs770830457 Chr.12: 96267179-96267179 on Build GRCh38
rs200030631 Chr.12: 96267043-96267043 on Build GRCh38
rs200976173 Chr.12: 96267074-96267074 on Build GRCh38
rs759096631 Chr.12: 96267083-96267084 on Build GRCh38
rs775680362 Chr.12: 96267078-96267078 on Build GRCh38
rs891438568 Chr.12: 96267038-96267038 on Build GRCh38
rs768697321 Chr.12: 96267089-96267089 on Build GRCh38
rs762757477 Chr.12: 96267090-96267090 on Build GRCh38
rs948640788 Chr.12: 96267188-96267188 on Build GRCh38
rs779351942 Chr.12: 96267132-96267132 on Build GRCh38
rs375758278 Chr.12: 96267202-96267202 on Build GRCh38

View Full Product Details