Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62603755-62604022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML3
Entrez Gene ID
256364
Gene Name
echinoderm microtubule associated protein like 3
Gene Aliases
ELP95
UniGene
Hs.379785
Gene Chromosome Location
Chr.11: 62602218-62613020 on Build GRCh38
Transcripts
NM_001300793.1 NM_001300794.1 NM_153265.2 XM_005273878.3 XM_006718489.3 XM_006718490.3 XM_006718491.3 XM_011544896.2 XM_011544897.2 XM_017017480.1 XM_017017481.1 XM_017017482.1 XM_017017483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929725 c.2192A>G p.H731R Chr.11: 62603794-62603794 on Build GRCh38
6581079 c.2152delC p.R718fs*45 Chr.11: 62603961-62603961 on Build GRCh38
3726587 c.2100T>C p.H700H Chr.11: 62604013-62604013 on Build GRCh38
6445677 c.2184C>T p.F728F Chr.11: 62603802-62603802 on Build GRCh38
4866559 c.2112C>T p.I704I Chr.11: 62604001-62604001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760729798 Chr.11: 62603793-62603794 on Build GRCh38
rs754261210 Chr.11: 62603920-62603920 on Build GRCh38
rs768437332 Chr.11: 62603894-62603894 on Build GRCh38
rs753293130 Chr.11: 62603935-62603935 on Build GRCh38
rs561089762 Chr.11: 62603938-62603938 on Build GRCh38
rs928390449 Chr.11: 62603928-62603928 on Build GRCh38
rs749852650 Chr.11: 62603959-62603959 on Build GRCh38
rs760041633 Chr.11: 62603922-62603922 on Build GRCh38
rs117319155 Chr.11: 62603901-62603901 on Build GRCh38
rs779613313 Chr.11: 62603972-62603972 on Build GRCh38
rs374644193 Chr.11: 62603902-62603902 on Build GRCh38
rs933123811 Chr.11: 62603895-62603895 on Build GRCh38
rs369199286 Chr.11: 62603852-62603852 on Build GRCh38
rs918882165 Chr.11: 62603939-62603939 on Build GRCh38
rs765999338 Chr.11: 62603824-62603824 on Build GRCh38
rs200929410 Chr.11: 62603861-62603861 on Build GRCh38
rs780785062 Chr.11: 62603950-62603950 on Build GRCh38
rs748928600 Chr.11: 62603977-62603977 on Build GRCh38
rs754033701 Chr.11: 62603967-62603967 on Build GRCh38
rs199877134 Chr.11: 62603985-62603985 on Build GRCh38
rs890566884 Chr.11: 62603816-62603816 on Build GRCh38
rs760983192 Chr.11: 62603912-62603912 on Build GRCh38
rs775285532 Chr.11: 62603859-62603859 on Build GRCh38
rs771858582 Chr.11: 62603988-62603988 on Build GRCh38
rs765680407 Chr.11: 62603923-62603923 on Build GRCh38
rs760314893 Chr.11: 62603780-62603780 on Build GRCh38
rs977545913 Chr.11: 62603914-62603914 on Build GRCh38
rs936593757 Chr.11: 62603860-62603860 on Build GRCh38
rs780541407 Chr.11: 62603793-62603793 on Build GRCh38
rs925229366 Chr.11: 62603832-62603832 on Build GRCh38
rs766653872 Chr.11: 62603918-62603918 on Build GRCh38
rs767675480 Chr.11: 62603905-62603905 on Build GRCh38
rs369013531 Chr.11: 62603961-62603961 on Build GRCh38
rs946044367 Chr.11: 62603822-62603822 on Build GRCh38
rs756846688 Chr.11: 62603786-62603786 on Build GRCh38
rs900336089 Chr.11: 62603893-62603894 on Build GRCh38
rs754732293 Chr.11: 62603909-62603909 on Build GRCh38
rs111813565 Chr.11: 62603881-62603881 on Build GRCh38

View Full Product Details