Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48703097-48703370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT2
Entrez Gene ID
2524
Gene Name
fucosyltransferase 2
Gene Aliases
B12QTL1, SE, SEC2, Se2, sej
UniGene
Hs.579928
Gene Chromosome Location
Chr.19: 48695971-48705934 on Build GRCh38
Transcripts
NM_000511.5 NM_001097638.2 NR_131188.1

Gene Symbol
LOC105447645
Entrez Gene ID
105447645
Gene Name
uncharacterized LOC105447645
Gene Aliases
-
UniGene
Hs.655052
Gene Chromosome Location
Chr.19: 48702450-48703848 on Build GRCh38
Transcripts
NM_000511.5 NM_001097638.2 NR_131188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712054 c.238G>A p.A80T Chr.19: 48703194-48703194 on Build GRCh38
1395087 c.249C>T p.Y83Y Chr.19: 48703205-48703205 on Build GRCh38
5081724 c.245_246delTG p.Y83fs*161 Chr.19: 48703201-48703202 on Build GRCh38
3757013 c.204A>G p.A68A Chr.19: 48703160-48703160 on Build GRCh38
4776796 c.155C>G p.S52* Chr.19: 48703111-48703111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs492602 Chr.19: 48703160-48703160 on Build GRCh38
rs768743754 Chr.19: 48703284-48703284 on Build GRCh38
rs994855043 Chr.19: 48703241-48703241 on Build GRCh38
rs764064106 Chr.19: 48703224-48703224 on Build GRCh38
rs548567519 Chr.19: 48703302-48703302 on Build GRCh38
rs534316263 Chr.19: 48703305-48703305 on Build GRCh38
rs755337838 Chr.19: 48703190-48703190 on Build GRCh38
rs935111256 Chr.19: 48703319-48703319 on Build GRCh38
rs143120145 Chr.19: 48703194-48703194 on Build GRCh38
rs756773151 Chr.19: 48703257-48703257 on Build GRCh38
rs575344194 Chr.19: 48703167-48703167 on Build GRCh38
rs760999180 Chr.19: 48703309-48703309 on Build GRCh38
rs281377 Chr.19: 48703346-48703346 on Build GRCh38
rs149356814 Chr.19: 48703267-48703267 on Build GRCh38
rs138507381 Chr.19: 48703201-48703201 on Build GRCh38
rs201211409 Chr.19: 48703188-48703188 on Build GRCh38
rs767931216 Chr.19: 48703243-48703243 on Build GRCh38
rs368563249 Chr.19: 48703193-48703193 on Build GRCh38
rs747300488 Chr.19: 48703158-48703167 on Build GRCh38
rs778485301 Chr.19: 48703261-48703261 on Build GRCh38
rs749842900 Chr.19: 48703325-48703325 on Build GRCh38
rs368598026 Chr.19: 48703341-48703341 on Build GRCh38
rs769066190 Chr.19: 48703123-48703123 on Build GRCh38
rs553851855 Chr.19: 48703210-48703210 on Build GRCh38
rs745525259 Chr.19: 48703262-48703262 on Build GRCh38
rs949095613 Chr.19: 48703185-48703185 on Build GRCh38
rs200375155 Chr.19: 48703310-48703310 on Build GRCh38
rs759340211 Chr.19: 48703159-48703159 on Build GRCh38
rs781695070 Chr.19: 48703129-48703129 on Build GRCh38
rs542733070 Chr.19: 48703169-48703169 on Build GRCh38
rs774040139 Chr.19: 48703155-48703155 on Build GRCh38
rs746750813 Chr.19: 48703279-48703279 on Build GRCh38
rs754326783 Chr.19: 48703311-48703311 on Build GRCh38
rs532253708 Chr.19: 48703251-48703251 on Build GRCh38
rs748750933 Chr.19: 48703198-48703198 on Build GRCh38
rs748945265 Chr.19: 48703146-48703146 on Build GRCh38
rs769281725 Chr.19: 48703136-48703136 on Build GRCh38
rs772918543 Chr.19: 48703142-48703142 on Build GRCh38
rs751793622 Chr.19: 48703232-48703232 on Build GRCh38
rs200868443 Chr.19: 48703292-48703292 on Build GRCh38
rs28362836 Chr.19: 48703304-48703304 on Build GRCh38
rs769531811 Chr.19: 48703267-48703269 on Build GRCh38
rs200157007 Chr.19: 48703291-48703291 on Build GRCh38
rs201304120 Chr.19: 48703187-48703187 on Build GRCh38
rs376091266 Chr.19: 48703245-48703245 on Build GRCh38
rs369690137 Chr.19: 48703293-48703293 on Build GRCh38
rs374416675 Chr.19: 48703168-48703168 on Build GRCh38
rs748840898 Chr.19: 48703253-48703253 on Build GRCh38
rs770562907 Chr.19: 48703151-48703151 on Build GRCh38
rs756780482 Chr.19: 48703313-48703313 on Build GRCh38
rs370354362 Chr.19: 48703228-48703228 on Build GRCh38
rs681343 Chr.19: 48703205-48703205 on Build GRCh38
rs370139701 Chr.19: 48703150-48703150 on Build GRCh38
rs199728421 Chr.19: 48703126-48703126 on Build GRCh38
rs112722916 Chr.19: 48703233-48703233 on Build GRCh38
rs776678286 Chr.19: 48703285-48703285 on Build GRCh38
rs890653259 Chr.19: 48703219-48703219 on Build GRCh38
rs142897699 Chr.19: 48703350-48703350 on Build GRCh38
rs771555785 Chr.19: 48703264-48703264 on Build GRCh38
rs771883357 Chr.19: 48703206-48703206 on Build GRCh38
rs201288465 Chr.19: 48703227-48703227 on Build GRCh38
rs747706140 Chr.19: 48703135-48703135 on Build GRCh38
rs770523746 Chr.19: 48703199-48703199 on Build GRCh38
rs571082951 Chr.19: 48703334-48703334 on Build GRCh38
rs138685530 Chr.19: 48703244-48703244 on Build GRCh38
rs746840651 Chr.19: 48703342-48703342 on Build GRCh38
rs778487951 Chr.19: 48703323-48703323 on Build GRCh38
rs758090404 Chr.19: 48703328-48703328 on Build GRCh38
rs760690963 Chr.19: 48703223-48703223 on Build GRCh38
rs762991501 Chr.19: 48703286-48703286 on Build GRCh38

View Full Product Details