Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144355456-144355725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5 NM_001280561.1

Gene Symbol
TMEM249
Entrez Gene ID
340393
Gene Name
transmembrane protein 249
Gene Aliases
C8ORFK29
UniGene
Hs.122231
Gene Chromosome Location
Chr.8: 144353226-144355609 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5 NM_001280561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683894 c.1505A>G p.Y502C Chr.8: 144355646-144355646 on Build GRCh38
1097204 c.1493C>T p.P498L Chr.8: 144355658-144355658 on Build GRCh38
3665321 c.1558C>T p.R520W Chr.8: 144355593-144355593 on Build GRCh38
3412854 c.1495G>C p.G499R Chr.8: 144355656-144355656 on Build GRCh38
5685015 c.1550G>T p.R517L Chr.8: 144355601-144355601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145097703 Chr.8: 144355592-144355592 on Build GRCh38
rs782537061 Chr.8: 144355568-144355568 on Build GRCh38
rs782355848 Chr.8: 144355689-144355689 on Build GRCh38
rs782049539 Chr.8: 144355610-144355610 on Build GRCh38
rs782689624 Chr.8: 144355644-144355644 on Build GRCh38
rs369046077 Chr.8: 144355494-144355494 on Build GRCh38
rs782075279 Chr.8: 144355654-144355655 on Build GRCh38
rs146434678 Chr.8: 144355534-144355534 on Build GRCh38
rs1030173449 Chr.8: 144355686-144355686 on Build GRCh38
rs782560261 Chr.8: 144355495-144355495 on Build GRCh38
rs782080344 Chr.8: 144355678-144355678 on Build GRCh38
rs150488922 Chr.8: 144355596-144355596 on Build GRCh38
rs967284791 Chr.8: 144355665-144355665 on Build GRCh38
rs141323222 Chr.8: 144355577-144355577 on Build GRCh38
rs782288059 Chr.8: 144355659-144355659 on Build GRCh38
rs781970557 Chr.8: 144355598-144355598 on Build GRCh38
rs781933763 Chr.8: 144355669-144355669 on Build GRCh38
rs534505390 Chr.8: 144355558-144355558 on Build GRCh38
rs976366789 Chr.8: 144355668-144355668 on Build GRCh38
rs781879109 Chr.8: 144355635-144355635 on Build GRCh38
rs782556212 Chr.8: 144355506-144355506 on Build GRCh38
rs782649430 Chr.8: 144355492-144355492 on Build GRCh38
rs781901139 Chr.8: 144355652-144355653 on Build GRCh38
rs200430164 Chr.8: 144355705-144355705 on Build GRCh38
rs149455097 Chr.8: 144355657-144355657 on Build GRCh38
rs782605034 Chr.8: 144355581-144355581 on Build GRCh38
rs781937338 Chr.8: 144355546-144355546 on Build GRCh38
rs782812176 Chr.8: 144355571-144355571 on Build GRCh38
rs782082942 Chr.8: 144355549-144355549 on Build GRCh38
rs782505176 Chr.8: 144355585-144355585 on Build GRCh38
rs782033960 Chr.8: 144355609-144355609 on Build GRCh38
rs538346478 Chr.8: 144355602-144355602 on Build GRCh38
rs782746141 Chr.8: 144355551-144355551 on Build GRCh38
rs782542949 Chr.8: 144355642-144355642 on Build GRCh38
rs782332270 Chr.8: 144355518-144355518 on Build GRCh38
rs151061162 Chr.8: 144355543-144355543 on Build GRCh38
rs782187820 Chr.8: 144355513-144355513 on Build GRCh38
rs782217946 Chr.8: 144355584-144355584 on Build GRCh38
rs782126106 Chr.8: 144355601-144355601 on Build GRCh38
rs781980576 Chr.8: 144355693-144355693 on Build GRCh38
rs140699050 Chr.8: 144355595-144355595 on Build GRCh38
rs781796083 Chr.8: 144355645-144355645 on Build GRCh38
rs561259699 Chr.8: 144355580-144355580 on Build GRCh38
rs782769335 Chr.8: 144355543-144355543 on Build GRCh38
rs138981805 Chr.8: 144355593-144355593 on Build GRCh38
rs377671954 Chr.8: 144355652-144355652 on Build GRCh38
rs561391486 Chr.8: 144355626-144355626 on Build GRCh38
rs782255518 Chr.8: 144355535-144355535 on Build GRCh38
rs1011236482 Chr.8: 144355639-144355639 on Build GRCh38
rs782397152 Chr.8: 144355540-144355540 on Build GRCh38

View Full Product Details