Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47576871-47577132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG5
Entrez Gene ID
10675
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
NGC
UniGene
Hs.45127
Gene Chromosome Location
Chr.3: 47562238-47580240 on Build GRCh38
Transcripts
NM_001206942.1 NM_001206943.1 NM_001206944.1 NM_001206945.1 NM_006574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316926 c.1072G>A p.E358K Chr.3: 47576954-47576954 on Build GRCh38
120051 c.1094G>A p.R365Q Chr.3: 47576932-47576932 on Build GRCh38
5630367 c.1071T>G p.T357T Chr.3: 47576955-47576955 on Build GRCh38
4812824 c.1123G>A p.D375N Chr.3: 47576903-47576903 on Build GRCh38
6226072 c.1040G>A p.G347D Chr.3: 47576986-47576986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141635864 Chr.3: 47577012-47577012 on Build GRCh38
rs781045312 Chr.3: 47576945-47576945 on Build GRCh38
rs773659783 Chr.3: 47577004-47577004 on Build GRCh38
rs748645809 Chr.3: 47577063-47577063 on Build GRCh38
rs891524399 Chr.3: 47576979-47576979 on Build GRCh38
rs146736749 Chr.3: 47576965-47576965 on Build GRCh38
rs751294641 Chr.3: 47577051-47577051 on Build GRCh38
rs964691566 Chr.3: 47577049-47577049 on Build GRCh38
rs754259161 Chr.3: 47576911-47576911 on Build GRCh38
rs777267541 Chr.3: 47577001-47577001 on Build GRCh38
rs761267592 Chr.3: 47576897-47576897 on Build GRCh38
rs775670678 Chr.3: 47576974-47576974 on Build GRCh38
rs746993899 Chr.3: 47576989-47576989 on Build GRCh38
rs770381855 Chr.3: 47577082-47577082 on Build GRCh38
rs373067610 Chr.3: 47577015-47577015 on Build GRCh38
rs750989919 Chr.3: 47576928-47576928 on Build GRCh38
rs147503336 Chr.3: 47576995-47576995 on Build GRCh38
rs566330604 Chr.3: 47576957-47576957 on Build GRCh38
rs983265672 Chr.3: 47577088-47577088 on Build GRCh38
rs979806637 Chr.3: 47577035-47577035 on Build GRCh38
rs765934635 Chr.3: 47576927-47576927 on Build GRCh38
rs773713041 Chr.3: 47577089-47577089 on Build GRCh38
rs201571337 Chr.3: 47577103-47577103 on Build GRCh38
rs142769230 Chr.3: 47577026-47577026 on Build GRCh38
rs199594550 Chr.3: 47577027-47577027 on Build GRCh38
rs752598799 Chr.3: 47577023-47577023 on Build GRCh38
rs751089835 Chr.3: 47577014-47577014 on Build GRCh38
rs773870725 Chr.3: 47577100-47577100 on Build GRCh38
rs765043397 Chr.3: 47576918-47576918 on Build GRCh38
rs755004660 Chr.3: 47577057-47577057 on Build GRCh38
rs138618029 Chr.3: 47576955-47576955 on Build GRCh38
rs772138256 Chr.3: 47576973-47576973 on Build GRCh38
rs376005436 Chr.3: 47577039-47577039 on Build GRCh38
rs555156953 Chr.3: 47577017-47577017 on Build GRCh38
rs780082287 Chr.3: 47577033-47577033 on Build GRCh38
rs765668169 Chr.3: 47577002-47577002 on Build GRCh38
rs368948134 Chr.3: 47576991-47576991 on Build GRCh38
rs201338850 Chr.3: 47576947-47576947 on Build GRCh38
rs762695503 Chr.3: 47577011-47577011 on Build GRCh38
rs147289456 Chr.3: 47577111-47577111 on Build GRCh38
rs762214843 Chr.3: 47576912-47576912 on Build GRCh38
rs764469635 Chr.3: 47576899-47576899 on Build GRCh38
rs757889508 Chr.3: 47576925-47576925 on Build GRCh38
rs1000311574 Chr.3: 47576948-47576948 on Build GRCh38
rs750098976 Chr.3: 47576920-47576920 on Build GRCh38
rs768618490 Chr.3: 47577046-47577046 on Build GRCh38
rs922413792 Chr.3: 47577101-47577101 on Build GRCh38
rs372995316 Chr.3: 47576933-47576933 on Build GRCh38

View Full Product Details