Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67611203-67611474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTCF
Entrez Gene ID
10664
Gene Name
CCCTC-binding factor
Gene Aliases
MRD21
UniGene
Hs.368367
Gene Chromosome Location
Chr.16: 67562407-67639185 on Build GRCh38
Transcripts
NM_001191022.1 NM_006565.3 XM_005255775.3 XM_017022868.1 XM_017022869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6845235 c.575_576insA p.D194fs*36 Chr.16: 67611407-67611408 on Build GRCh38
6845266 c.383delC p.T129fs*3 Chr.16: 67611215-67611215 on Build GRCh38
2995127 c.552_553insC p.Q186fs*5 Chr.16: 67611384-67611385 on Build GRCh38
6974544 c.430A>T p.K144* Chr.16: 67611262-67611262 on Build GRCh38
6915704 c.615G>T p.K205N Chr.16: 67611447-67611447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369939556 Chr.16: 67611237-67611237 on Build GRCh38
rs763743813 Chr.16: 67611330-67611330 on Build GRCh38
rs373823720 Chr.16: 67611247-67611247 on Build GRCh38
rs779020931 Chr.16: 67611268-67611268 on Build GRCh38
rs377709753 Chr.16: 67611277-67611277 on Build GRCh38
rs866170521 Chr.16: 67611443-67611443 on Build GRCh38
rs758670950 Chr.16: 67611433-67611433 on Build GRCh38
rs759449563 Chr.16: 67611285-67611285 on Build GRCh38
rs756610479 Chr.16: 67611387-67611387 on Build GRCh38
rs749650039 Chr.16: 67611248-67611248 on Build GRCh38
rs779404795 Chr.16: 67611408-67611408 on Build GRCh38
rs376744931 Chr.16: 67611312-67611312 on Build GRCh38
rs761761096 Chr.16: 67611430-67611430 on Build GRCh38
rs754178156 Chr.16: 67611398-67611398 on Build GRCh38
rs933470800 Chr.16: 67611271-67611271 on Build GRCh38
rs200516339 Chr.16: 67611228-67611228 on Build GRCh38
rs767120755 Chr.16: 67611360-67611360 on Build GRCh38
rs773340288 Chr.16: 67611279-67611279 on Build GRCh38
rs769661363 Chr.16: 67611306-67611306 on Build GRCh38
rs749999010 Chr.16: 67611376-67611376 on Build GRCh38
rs748358750 Chr.16: 67611428-67611428 on Build GRCh38
rs148372058 Chr.16: 67611276-67611276 on Build GRCh38
rs373511929 Chr.16: 67611280-67611280 on Build GRCh38
rs774149823 Chr.16: 67611342-67611342 on Build GRCh38
rs766823310 Chr.16: 67611389-67611389 on Build GRCh38
rs750959177 Chr.16: 67611230-67611230 on Build GRCh38
rs762678010 Chr.16: 67611321-67611321 on Build GRCh38
rs768877983 Chr.16: 67611259-67611259 on Build GRCh38
rs878885019 Chr.16: 67611418-67611418 on Build GRCh38
rs886041997 Chr.16: 67611442-67611443 on Build GRCh38
rs748162324 Chr.16: 67611269-67611269 on Build GRCh38
rs761623880 Chr.16: 67611347-67611347 on Build GRCh38

View Full Product Details