Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32226302-32226559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3I
Entrez Gene ID
8668
Gene Name
eukaryotic translation initiation factor 3 subunit I
Gene Aliases
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36
UniGene
Hs.530096
Gene Chromosome Location
Chr.1: 32221584-32231604 on Build GRCh38
Transcripts
NM_003757.3 XM_017002671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4970246 c.401-4A>G p.? Chr.1: 32226399-32226399 on Build GRCh38
4846058 c.503G>A p.G168E Chr.1: 32226505-32226505 on Build GRCh38
3977584 c.387T>C p.D129D Chr.1: 32226307-32226307 on Build GRCh38
1296291 c.505G>C p.E169Q Chr.1: 32226507-32226507 on Build GRCh38
1296290 c.396G>C p.Q132H Chr.1: 32226316-32226316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751405254 Chr.1: 32226357-32226357 on Build GRCh38
rs965760369 Chr.1: 32226327-32226327 on Build GRCh38
rs76933339 Chr.1: 32226345-32226345 on Build GRCh38
rs572553090 Chr.1: 32226358-32226358 on Build GRCh38
rs780279891 Chr.1: 32226536-32226536 on Build GRCh38
rs780478406 Chr.1: 32226414-32226414 on Build GRCh38
rs774203133 Chr.1: 32226476-32226476 on Build GRCh38
rs758637679 Chr.1: 32226410-32226410 on Build GRCh38
rs750551140 Chr.1: 32226514-32226514 on Build GRCh38
rs758647572 Chr.1: 32226527-32226527 on Build GRCh38
rs111982592 Chr.1: 32226322-32226322 on Build GRCh38
rs200845980 Chr.1: 32226479-32226479 on Build GRCh38
rs754027887 Chr.1: 32226371-32226371 on Build GRCh38
rs761452536 Chr.1: 32226339-32226339 on Build GRCh38
rs113367121 Chr.1: 32226321-32226321 on Build GRCh38
rs201776715 Chr.1: 32226395-32226395 on Build GRCh38
rs765426821 Chr.1: 32226506-32226506 on Build GRCh38
rs759377657 Chr.1: 32226486-32226486 on Build GRCh38
rs769222429 Chr.1: 32226420-32226420 on Build GRCh38
rs769408795 Chr.1: 32226459-32226459 on Build GRCh38
rs377229890 Chr.1: 32226361-32226361 on Build GRCh38
rs866279978 Chr.1: 32226481-32226481 on Build GRCh38
rs772972113 Chr.1: 32226343-32226343 on Build GRCh38
rs529692171 Chr.1: 32226488-32226488 on Build GRCh38
rs777766050 Chr.1: 32226539-32226539 on Build GRCh38
rs1042772235 Chr.1: 32226390-32226390 on Build GRCh38
rs780669264 Chr.1: 32226426-32226426 on Build GRCh38
rs573555723 Chr.1: 32226436-32226436 on Build GRCh38
rs114114874 Chr.1: 32226368-32226368 on Build GRCh38
rs776125853 Chr.1: 32226323-32226323 on Build GRCh38
rs772813256 Chr.1: 32226466-32226466 on Build GRCh38
rs766228235 Chr.1: 32226347-32226347 on Build GRCh38
rs764932109 Chr.1: 32226342-32226342 on Build GRCh38
rs772912286 Chr.1: 32226375-32226375 on Build GRCh38
rs769246019 Chr.1: 32226372-32226372 on Build GRCh38
rs902971917 Chr.1: 32226404-32226404 on Build GRCh38

View Full Product Details