Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 26994825-26995066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBIN
Entrez Gene ID
387758
Gene Name
fin bud initiation factor homolog (zebrafish)
Gene Aliases
-
UniGene
Hs.712718
Gene Chromosome Location
Chr.11: 26994081-26997087 on Build GRCh38
Transcripts
NM_203371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617898 c.338A>G p.E113G Chr.11: 26994864-26994864 on Build GRCh38
5728378 c.529C>T p.L177L Chr.11: 26995055-26995055 on Build GRCh38
5661409 c.465G>C p.E155D Chr.11: 26994991-26994991 on Build GRCh38
5372309 c.381G>A p.G127G Chr.11: 26994907-26994907 on Build GRCh38
2113250 c.417G>A p.E139E Chr.11: 26994943-26994943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754490539 Chr.11: 26994959-26994959 on Build GRCh38
rs753777074 Chr.11: 26994862-26994862 on Build GRCh38
rs767662457 Chr.11: 26994917-26994917 on Build GRCh38
rs765208032 Chr.11: 26994932-26994932 on Build GRCh38
rs771095824 Chr.11: 26994907-26994907 on Build GRCh38
rs200720522 Chr.11: 26994954-26994954 on Build GRCh38
rs746550163 Chr.11: 26994885-26994885 on Build GRCh38
rs562967725 Chr.11: 26994987-26994987 on Build GRCh38
rs768108138 Chr.11: 26994890-26994890 on Build GRCh38
rs751228544 Chr.11: 26995021-26995021 on Build GRCh38
rs759709640 Chr.11: 26994915-26994915 on Build GRCh38
rs750251302 Chr.11: 26994934-26994934 on Build GRCh38
rs936582341 Chr.11: 26994914-26994914 on Build GRCh38
rs772290499 Chr.11: 26994985-26994985 on Build GRCh38
rs367758269 Chr.11: 26994984-26994984 on Build GRCh38
rs751653174 Chr.11: 26994855-26994855 on Build GRCh38
rs185352306 Chr.11: 26994998-26994998 on Build GRCh38
rs765137980 Chr.11: 26995000-26995000 on Build GRCh38
rs762689940 Chr.11: 26995009-26995009 on Build GRCh38
rs144960366 Chr.11: 26994955-26994955 on Build GRCh38
rs145383958 Chr.11: 26994871-26994871 on Build GRCh38
rs774606887 Chr.11: 26994908-26994908 on Build GRCh38
rs975092793 Chr.11: 26994928-26994928 on Build GRCh38
rs199736217 Chr.11: 26994956-26994956 on Build GRCh38
rs759764078 Chr.11: 26994983-26994983 on Build GRCh38
rs144294030 Chr.11: 26994980-26994980 on Build GRCh38
rs368516233 Chr.11: 26994847-26994847 on Build GRCh38
rs747647368 Chr.11: 26994962-26994962 on Build GRCh38
rs141428961 Chr.11: 26994900-26994900 on Build GRCh38
rs781688110 Chr.11: 26994899-26994899 on Build GRCh38
rs767157136 Chr.11: 26995032-26995032 on Build GRCh38
rs917676485 Chr.11: 26994881-26994881 on Build GRCh38
rs774465844 Chr.11: 26994982-26994982 on Build GRCh38
rs764174247 Chr.11: 26994858-26994858 on Build GRCh38
rs149580940 Chr.11: 26994864-26994864 on Build GRCh38
rs540597862 Chr.11: 26994921-26994921 on Build GRCh38
rs372777320 Chr.11: 26994905-26994905 on Build GRCh38
rs778308233 Chr.11: 26994967-26994967 on Build GRCh38
rs140574615 Chr.11: 26994857-26994857 on Build GRCh38
rs780626122 Chr.11: 26994961-26994961 on Build GRCh38
rs902469109 Chr.11: 26994888-26994888 on Build GRCh38
rs774074818 Chr.11: 26994944-26994945 on Build GRCh38
rs372108528 Chr.11: 26994865-26994865 on Build GRCh38
rs181248433 Chr.11: 26994970-26994970 on Build GRCh38
rs773153187 Chr.11: 26995003-26995003 on Build GRCh38
rs754685334 Chr.11: 26995025-26995025 on Build GRCh38
rs758043137 Chr.11: 26994883-26994883 on Build GRCh38
rs753830629 Chr.11: 26994919-26994919 on Build GRCh38
rs779727131 Chr.11: 26994884-26994884 on Build GRCh38
rs767097250 Chr.11: 26994966-26994966 on Build GRCh38
rs552386801 Chr.11: 26995012-26995012 on Build GRCh38
rs750103673 Chr.11: 26994878-26994878 on Build GRCh38
rs770159379 Chr.11: 26994902-26994902 on Build GRCh38

View Full Product Details