Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99849326-99849581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMSS1
Entrez Gene ID
84319
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
C3orf26
UniGene
Hs.581320
Gene Chromosome Location
Chr.3: 99817834-100178632 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
UniGene
Hs.104672
Gene Chromosome Location
Chr.3: 99833144-100114513 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1131323 c.2225A>C p.D742A Chr.3: 99849451-99849451 on Build GRCh38
7206765 c.2230T>G p.S744A Chr.3: 99849446-99849446 on Build GRCh38
5636443 c.2349T>G p.P783P Chr.3: 99849327-99849327 on Build GRCh38
1207156 c.2166A>C p.L722F Chr.3: 99849510-99849510 on Build GRCh38
4328356 c.2240A>C p.Q747P Chr.3: 99849436-99849436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762729634 Chr.3: 99849455-99849455 on Build GRCh38
rs369205692 Chr.3: 99849516-99849516 on Build GRCh38
rs749879867 Chr.3: 99849494-99849494 on Build GRCh38
rs142569410 Chr.3: 99849488-99849488 on Build GRCh38
rs754312407 Chr.3: 99849512-99849512 on Build GRCh38
rs777954720 Chr.3: 99849542-99849542 on Build GRCh38
rs746318060 Chr.3: 99849352-99849352 on Build GRCh38
rs776467626 Chr.3: 99849420-99849420 on Build GRCh38
rs559963545 Chr.3: 99849451-99849451 on Build GRCh38
rs915342360 Chr.3: 99849515-99849515 on Build GRCh38
rs747026428 Chr.3: 99849544-99849544 on Build GRCh38
rs200655671 Chr.3: 99849517-99849517 on Build GRCh38
rs965955142 Chr.3: 99849370-99849370 on Build GRCh38
rs780420744 Chr.3: 99849501-99849501 on Build GRCh38
rs765012473 Chr.3: 99849548-99849550 on Build GRCh38
rs774702238 Chr.3: 99849404-99849404 on Build GRCh38
rs199915987 Chr.3: 99849550-99849550 on Build GRCh38
rs13353487 Chr.3: 99849441-99849441 on Build GRCh38
rs376645916 Chr.3: 99849419-99849419 on Build GRCh38
rs749900665 Chr.3: 99849547-99849548 on Build GRCh38
rs765060736 Chr.3: 99849450-99849450 on Build GRCh38
rs199518119 Chr.3: 99849395-99849395 on Build GRCh38
rs374837208 Chr.3: 99849499-99849499 on Build GRCh38
rs748255489 Chr.3: 99849413-99849413 on Build GRCh38
rs772070838 Chr.3: 99849538-99849538 on Build GRCh38
rs748343292 Chr.3: 99849518-99849518 on Build GRCh38
rs200062458 Chr.3: 99849551-99849551 on Build GRCh38
rs757846241 Chr.3: 99849445-99849445 on Build GRCh38
rs763793326 Chr.3: 99849487-99849487 on Build GRCh38
rs202066216 Chr.3: 99849385-99849385 on Build GRCh38

View Full Product Details