Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9443378-9443607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
UniGene
Hs.515972
Gene Chromosome Location
Chr.2: 9423537-9473110 on Build GRCh38
Transcripts
NM_001321833.1 NM_001321834.1 NM_001321835.1 NM_001321836.1 NM_016207.3

SNP Detail

SNP ID Chromosome Location
rs768086759 Chr.2: 9443500-9443500 on Build GRCh38
rs200713954 Chr.2: 9443559-9443559 on Build GRCh38
rs769436422 Chr.2: 9443467-9443467 on Build GRCh38
rs757617467 Chr.2: 9443529-9443529 on Build GRCh38
rs778361147 Chr.2: 9443487-9443495 on Build GRCh38
rs534335251 Chr.2: 9443485-9443485 on Build GRCh38
rs1041277998 Chr.2: 9443516-9443516 on Build GRCh38
rs909680577 Chr.2: 9443479-9443479 on Build GRCh38
rs756520531 Chr.2: 9443504-9443504 on Build GRCh38
rs773920802 Chr.2: 9443494-9443494 on Build GRCh38
rs776375392 Chr.2: 9443464-9443464 on Build GRCh38
rs747979949 Chr.2: 9443466-9443466 on Build GRCh38
rs137925570 Chr.2: 9443562-9443562 on Build GRCh38
rs760117150 Chr.2: 9443450-9443450 on Build GRCh38
rs371721884 Chr.2: 9443539-9443539 on Build GRCh38
rs79545197 Chr.2: 9443563-9443563 on Build GRCh38
rs779263224 Chr.2: 9443530-9443530 on Build GRCh38
rs984207704 Chr.2: 9443415-9443415 on Build GRCh38
rs1044067017 Chr.2: 9443577-9443577 on Build GRCh38
rs905530660 Chr.2: 9443579-9443579 on Build GRCh38
rs764599589 Chr.2: 9443508-9443508 on Build GRCh38
rs758343374 Chr.2: 9443556-9443556 on Build GRCh38
rs754222084 Chr.2: 9443525-9443525 on Build GRCh38
rs765987097 Chr.2: 9443491-9443491 on Build GRCh38
rs762645850 Chr.2: 9443489-9443489 on Build GRCh38
rs758960084 Chr.2: 9443495-9443495 on Build GRCh38
rs753100683 Chr.2: 9443503-9443503 on Build GRCh38
rs889775028 Chr.2: 9443442-9443443 on Build GRCh38

View Full Product Details