Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86611595-86611820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
UniGene
Hs.154433
Gene Chromosome Location
Chr.8: 86573608-86743675 on Build GRCh38
Transcripts
NM_019098.4 XM_011517138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955620 c.1633T>C p.Y545H Chr.8: 86611617-86611617 on Build GRCh38
7252479 c.1601A>G p.Y534C Chr.8: 86611649-86611649 on Build GRCh38
6323988 c.1583G>A p.C528Y Chr.8: 86611667-86611667 on Build GRCh38
6365854 c.1627G>T p.V543F Chr.8: 86611623-86611623 on Build GRCh38
6364658 c.1621A>G p.K541E Chr.8: 86611629-86611629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761000282 Chr.8: 86611712-86611712 on Build GRCh38
rs950991226 Chr.8: 86611773-86611773 on Build GRCh38
rs1025166276 Chr.8: 86611759-86611759 on Build GRCh38
rs750366014 Chr.8: 86611699-86611699 on Build GRCh38
rs770018514 Chr.8: 86611676-86611676 on Build GRCh38
rs114087363 Chr.8: 86611752-86611752 on Build GRCh38
rs769226980 Chr.8: 86611652-86611652 on Build GRCh38
rs746817996 Chr.8: 86611623-86611623 on Build GRCh38
rs895314547 Chr.8: 86611721-86611721 on Build GRCh38
rs1013245104 Chr.8: 86611764-86611764 on Build GRCh38
rs35903042 Chr.8: 86611624-86611624 on Build GRCh38
rs549945596 Chr.8: 86611768-86611768 on Build GRCh38
rs1035474610 Chr.8: 86611734-86611734 on Build GRCh38
rs1057516504 Chr.8: 86611672-86611672 on Build GRCh38
rs747653530 Chr.8: 86611628-86611628 on Build GRCh38
rs367652647 Chr.8: 86611643-86611643 on Build GRCh38
rs903872128 Chr.8: 86611671-86611671 on Build GRCh38
rs770523874 Chr.8: 86611694-86611694 on Build GRCh38
rs772725807 Chr.8: 86611673-86611673 on Build GRCh38
rs775895294 Chr.8: 86611709-86611709 on Build GRCh38
rs762621879 Chr.8: 86611684-86611684 on Build GRCh38
rs781166193 Chr.8: 86611627-86611627 on Build GRCh38
rs377603878 Chr.8: 86611636-86611636 on Build GRCh38

View Full Product Details