Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6977150-6977395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NR_135131.1 NM_005768.5

Gene Symbol
LPCAT3
Entrez Gene ID
10162
Gene Name
lysophosphatidylcholine acyltransferase 3
Gene Aliases
C3F, LPCAT, LPLAT 5, LPSAT, MBOAT5, OACT5, nessy
UniGene
Hs.655248
Gene Chromosome Location
Chr.12: 6976185-7018538 on Build GRCh38
Transcripts
NR_135131.1 NM_005768.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180827 c.1436G>T p.R479M Chr.12: 6977174-6977174 on Build GRCh38
5621164 c.1373G>C p.G458A Chr.12: 6977237-6977237 on Build GRCh38
468863 c.1381T>A p.F461I Chr.12: 6977229-6977229 on Build GRCh38
6702325 c.1454_1456delAGA p.K485delK Chr.12: 6977154-6977156 on Build GRCh38
4430938 c.1404A>G p.I468M Chr.12: 6977206-6977206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781832108 Chr.12: 6977252-6977252 on Build GRCh38
rs147741167 Chr.12: 6977181-6977181 on Build GRCh38
rs782491517 Chr.12: 6977217-6977217 on Build GRCh38
rs782117795 Chr.12: 6977345-6977345 on Build GRCh38
rs782057653 Chr.12: 6977202-6977202 on Build GRCh38
rs781810040 Chr.12: 6977229-6977229 on Build GRCh38
rs781908784 Chr.12: 6977199-6977199 on Build GRCh38
rs782045785 Chr.12: 6977321-6977321 on Build GRCh38
rs782718258 Chr.12: 6977208-6977208 on Build GRCh38
rs782122398 Chr.12: 6977276-6977276 on Build GRCh38
rs756685907 Chr.12: 6977228-6977228 on Build GRCh38
rs782437751 Chr.12: 6977245-6977245 on Build GRCh38
rs782273703 Chr.12: 6977257-6977257 on Build GRCh38
rs782801224 Chr.12: 6977184-6977184 on Build GRCh38
rs142552475 Chr.12: 6977367-6977367 on Build GRCh38
rs111742644 Chr.12: 6977369-6977369 on Build GRCh38
rs921327898 Chr.12: 6977231-6977231 on Build GRCh38
rs778625660 Chr.12: 6977342-6977342 on Build GRCh38
rs782623914 Chr.12: 6977273-6977273 on Build GRCh38
rs78821515 Chr.12: 6977317-6977317 on Build GRCh38
rs781931181 Chr.12: 6977305-6977305 on Build GRCh38
rs782393457 Chr.12: 6977269-6977269 on Build GRCh38
rs782341445 Chr.12: 6977300-6977300 on Build GRCh38
rs371126597 Chr.12: 6977223-6977223 on Build GRCh38
rs782416066 Chr.12: 6977341-6977341 on Build GRCh38
rs782685375 Chr.12: 6977246-6977246 on Build GRCh38
rs781860896 Chr.12: 6977212-6977212 on Build GRCh38

View Full Product Details