Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46777515-46777774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESD
Entrez Gene ID
2098
Gene Name
esterase D
Gene Aliases
FGH
UniGene
Hs.432491
Gene Chromosome Location
Chr.13: 46771256-46797681 on Build GRCh38
Transcripts
NM_001984.1 XM_005266278.2 XM_011534954.1 XM_017020414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047905 c.638G>T p.G213V Chr.13: 46777586-46777586 on Build GRCh38
1367093 c.640T>G p.S214A Chr.13: 46777584-46777584 on Build GRCh38
4454935 c.617A>T p.H206L Chr.13: 46777607-46777607 on Build GRCh38
6876754 c.685C>T p.L229F Chr.13: 46777539-46777539 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs916677670 Chr.13: 46777588-46777588 on Build GRCh38
rs779095177 Chr.13: 46777621-46777621 on Build GRCh38
rs775919961 Chr.13: 46777712-46777712 on Build GRCh38
rs779585231 Chr.13: 46777605-46777605 on Build GRCh38
rs759443898 Chr.13: 46777613-46777614 on Build GRCh38
rs376230652 Chr.13: 46777549-46777549 on Build GRCh38
rs563977652 Chr.13: 46777694-46777694 on Build GRCh38
rs771446760 Chr.13: 46777653-46777653 on Build GRCh38
rs985400919 Chr.13: 46777686-46777686 on Build GRCh38
rs888888947 Chr.13: 46777714-46777714 on Build GRCh38
rs1008647147 Chr.13: 46777716-46777716 on Build GRCh38
rs369240543 Chr.13: 46777625-46777625 on Build GRCh38
rs918799369 Chr.13: 46777555-46777555 on Build GRCh38
rs746319600 Chr.13: 46777645-46777645 on Build GRCh38
rs780955943 Chr.13: 46777655-46777655 on Build GRCh38
rs754090039 Chr.13: 46777607-46777607 on Build GRCh38
rs769382359 Chr.13: 46777551-46777551 on Build GRCh38
rs199632443 Chr.13: 46777556-46777556 on Build GRCh38
rs767410255 Chr.13: 46777620-46777622 on Build GRCh38
rs1020234876 Chr.13: 46777720-46777720 on Build GRCh38
rs372869959 Chr.13: 46777674-46777674 on Build GRCh38
rs771094740 Chr.13: 46777585-46777585 on Build GRCh38
rs775124226 Chr.13: 46777661-46777661 on Build GRCh38
rs762996843 Chr.13: 46777574-46777574 on Build GRCh38
rs980470615 Chr.13: 46777638-46777638 on Build GRCh38
rs751424461 Chr.13: 46777580-46777580 on Build GRCh38
rs754981057 Chr.13: 46777618-46777618 on Build GRCh38
rs753127193 Chr.13: 46777623-46777623 on Build GRCh38
rs868080158 Chr.13: 46777596-46777596 on Build GRCh38
rs906358890 Chr.13: 46777636-46777636 on Build GRCh38
rs778427043 Chr.13: 46777635-46777635 on Build GRCh38
rs940928811 Chr.13: 46777700-46777700 on Build GRCh38
rs763785998 Chr.13: 46777577-46777577 on Build GRCh38
rs965581569 Chr.13: 46777724-46777724 on Build GRCh38
rs747323190 Chr.13: 46777637-46777637 on Build GRCh38
rs761433242 Chr.13: 46777742-46777742 on Build GRCh38
rs777355329 Chr.13: 46777665-46777666 on Build GRCh38
rs769166333 Chr.13: 46777660-46777660 on Build GRCh38
rs755511121 Chr.13: 46777654-46777655 on Build GRCh38
rs761608369 Chr.13: 46777595-46777595 on Build GRCh38
rs745578218 Chr.13: 46777659-46777659 on Build GRCh38
rs752246538 Chr.13: 46777631-46777635 on Build GRCh38

View Full Product Details