Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35130233-35130506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC8
Entrez Gene ID
54752
Gene Name
fibronectin type III domain containing 8
Gene Aliases
-
UniGene
Hs.120369
Gene Chromosome Location
Chr.17: 35121612-35130732 on Build GRCh38
Transcripts
NM_017559.2 XM_017024777.1

Gene Symbol
NLE1
Entrez Gene ID
54475
Gene Name
notchless homolog 1
Gene Aliases
Nle
UniGene
Hs.85570
Gene Chromosome Location
Chr.17: 35128755-35142315 on Build GRCh38
Transcripts
NM_017559.2 XM_017024777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6523440 c.946G>A p.E316K Chr.17: 35130405-35130405 on Build GRCh38
3516174 c.923G>A p.G308E Chr.17: 35130382-35130382 on Build GRCh38
1382406 c.901C>T p.R301W Chr.17: 35130360-35130360 on Build GRCh38
5930337 c.949G>C p.D317H Chr.17: 35130408-35130408 on Build GRCh38
3516173 c.876G>A p.Q292Q Chr.17: 35130335-35130335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202047426 Chr.17: 35130349-35130349 on Build GRCh38
rs771114524 Chr.17: 35130262-35130263 on Build GRCh38
rs761893208 Chr.17: 35130272-35130272 on Build GRCh38
rs754322331 Chr.17: 35130486-35130486 on Build GRCh38
rs753511517 Chr.17: 35130278-35130278 on Build GRCh38
rs144708359 Chr.17: 35130380-35130380 on Build GRCh38
rs141830915 Chr.17: 35130436-35130436 on Build GRCh38
rs915792745 Chr.17: 35130353-35130353 on Build GRCh38
rs770419514 Chr.17: 35130337-35130337 on Build GRCh38
rs557687756 Chr.17: 35130344-35130344 on Build GRCh38
rs531208768 Chr.17: 35130339-35130339 on Build GRCh38
rs769220051 Chr.17: 35130383-35130383 on Build GRCh38
rs1018510152 Chr.17: 35130435-35130435 on Build GRCh38
rs754801014 Chr.17: 35130289-35130289 on Build GRCh38
rs752598812 Chr.17: 35130300-35130300 on Build GRCh38
rs148514334 Chr.17: 35130398-35130398 on Build GRCh38
rs199634203 Chr.17: 35130396-35130396 on Build GRCh38
rs750125681 Chr.17: 35130452-35130452 on Build GRCh38
rs773336985 Chr.17: 35130397-35130397 on Build GRCh38
rs751842705 Chr.17: 35130264-35130264 on Build GRCh38
rs774238390 Chr.17: 35130345-35130345 on Build GRCh38
rs146757908 Chr.17: 35130341-35130341 on Build GRCh38
rs199607691 Chr.17: 35130342-35130342 on Build GRCh38
rs1007083520 Chr.17: 35130423-35130423 on Build GRCh38
rs762728015 Chr.17: 35130411-35130411 on Build GRCh38
rs780753986 Chr.17: 35130378-35130378 on Build GRCh38
rs774625655 Chr.17: 35130265-35130267 on Build GRCh38
rs780571323 Chr.17: 35130482-35130482 on Build GRCh38
rs966750957 Chr.17: 35130317-35130317 on Build GRCh38
rs756759890 Chr.17: 35130444-35130444 on Build GRCh38
rs537116877 Chr.17: 35130360-35130360 on Build GRCh38
rs755774636 Chr.17: 35130460-35130460 on Build GRCh38
rs373790485 Chr.17: 35130402-35130402 on Build GRCh38
rs751094213 Chr.17: 35130372-35130372 on Build GRCh38
rs372341617 Chr.17: 35130384-35130384 on Build GRCh38
rs147465242 Chr.17: 35130361-35130361 on Build GRCh38
rs78526886 Chr.17: 35130297-35130297 on Build GRCh38
rs149380490 Chr.17: 35130371-35130371 on Build GRCh38
rs767016976 Chr.17: 35130450-35130450 on Build GRCh38
rs374433104 Chr.17: 35130348-35130348 on Build GRCh38
rs371002417 Chr.17: 35130298-35130298 on Build GRCh38
rs777215406 Chr.17: 35130315-35130315 on Build GRCh38
rs748236400 Chr.17: 35130386-35130386 on Build GRCh38
rs746683874 Chr.17: 35130322-35130322 on Build GRCh38
rs373413395 Chr.17: 35130262-35130262 on Build GRCh38
rs777193469 Chr.17: 35130441-35130441 on Build GRCh38
rs979289115 Chr.17: 35130269-35130269 on Build GRCh38
rs763993000 Chr.17: 35130424-35130424 on Build GRCh38
rs144023197 Chr.17: 35130290-35130290 on Build GRCh38
rs543199253 Chr.17: 35130406-35130406 on Build GRCh38
rs756775303 Chr.17: 35130375-35130375 on Build GRCh38
rs762100704 Chr.17: 35130347-35130347 on Build GRCh38
rs957264103 Chr.17: 35130346-35130346 on Build GRCh38
rs757933763 Chr.17: 35130301-35130301 on Build GRCh38
rs370230473 Chr.17: 35130381-35130381 on Build GRCh38
rs766211635 Chr.17: 35130275-35130275 on Build GRCh38

View Full Product Details