Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65870122-65870347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP2
Entrez Gene ID
30008
Gene Name
EGF containing fibulin like extracellular matrix protein 2
Gene Aliases
ARCL1B, FBLN4, MBP1, UPH1
UniGene
Hs.731454
Gene Chromosome Location
Chr.11: 65866441-65872934 on Build GRCh38
Transcripts
NM_016938.4 NR_037718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384425 c.492C>A p.D164E Chr.11: 65870236-65870236 on Build GRCh38
930530 c.596G>A p.R199H Chr.11: 65870132-65870132 on Build GRCh38
5839277 c.583G>A p.G195R Chr.11: 65870145-65870145 on Build GRCh38
6306864 c.506G>T p.R169L Chr.11: 65870222-65870222 on Build GRCh38
1628170 c.595C>A p.R199S Chr.11: 65870133-65870133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143662598 Chr.11: 65870174-65870174 on Build GRCh38
rs752456997 Chr.11: 65870142-65870142 on Build GRCh38
rs374413025 Chr.11: 65870283-65870283 on Build GRCh38
rs145853251 Chr.11: 65870158-65870158 on Build GRCh38
rs773297046 Chr.11: 65870202-65870202 on Build GRCh38
rs991647691 Chr.11: 65870301-65870301 on Build GRCh38
rs775164905 Chr.11: 65870175-65870175 on Build GRCh38
rs148940436 Chr.11: 65870163-65870163 on Build GRCh38
rs1043355 Chr.11: 65870155-65870155 on Build GRCh38
rs942484558 Chr.11: 65870243-65870243 on Build GRCh38
rs193302870 Chr.11: 65870151-65870151 on Build GRCh38
rs147124207 Chr.11: 65870302-65870302 on Build GRCh38
rs762712612 Chr.11: 65870223-65870223 on Build GRCh38
rs781247249 Chr.11: 65870255-65870257 on Build GRCh38
rs35726936 Chr.11: 65870227-65870228 on Build GRCh38
rs570927073 Chr.11: 65870164-65870164 on Build GRCh38
rs564935293 Chr.11: 65870273-65870273 on Build GRCh38
rs769756316 Chr.11: 65870258-65870259 on Build GRCh38
rs934226469 Chr.11: 65870255-65870255 on Build GRCh38
rs749980581 Chr.11: 65870242-65870242 on Build GRCh38
rs749651807 Chr.11: 65870182-65870182 on Build GRCh38
rs570696654 Chr.11: 65870266-65870267 on Build GRCh38
rs367645893 Chr.11: 65870252-65870252 on Build GRCh38
rs772504760 Chr.11: 65870201-65870201 on Build GRCh38
rs937096955 Chr.11: 65870297-65870297 on Build GRCh38
rs200648492 Chr.11: 65870265-65870265 on Build GRCh38
rs763944898 Chr.11: 65870229-65870229 on Build GRCh38
rs769039425 Chr.11: 65870191-65870191 on Build GRCh38
rs149021732 Chr.11: 65870259-65870265 on Build GRCh38
rs757273990 Chr.11: 65870230-65870230 on Build GRCh38
rs562466098 Chr.11: 65870159-65870159 on Build GRCh38
rs148075092 Chr.11: 65870217-65870217 on Build GRCh38
rs779343553 Chr.11: 65870270-65870270 on Build GRCh38
rs755651962 Chr.11: 65870248-65870248 on Build GRCh38
rs780195096 Chr.11: 65870259-65870260 on Build GRCh38
rs1053118642 Chr.11: 65870296-65870296 on Build GRCh38
rs762305922 Chr.11: 65870197-65870197 on Build GRCh38
rs760696788 Chr.11: 65870216-65870216 on Build GRCh38
rs972385473 Chr.11: 65870208-65870208 on Build GRCh38
rs928350405 Chr.11: 65870278-65870278 on Build GRCh38
rs774781166 Chr.11: 65870196-65870196 on Build GRCh38
rs145037272 Chr.11: 65870232-65870232 on Build GRCh38
rs370392978 Chr.11: 65870276-65870276 on Build GRCh38
rs758174325 Chr.11: 65870146-65870146 on Build GRCh38
rs768643897 Chr.11: 65870267-65870273 on Build GRCh38
rs779619124 Chr.11: 65870254-65870254 on Build GRCh38
rs141310608 Chr.11: 65870222-65870222 on Build GRCh38
rs748959074 Chr.11: 65870263-65870263 on Build GRCh38

View Full Product Details