Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233438244-233438487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
UniGene
Hs.471675
Gene Chromosome Location
Chr.2: 233354507-233472098 on Build GRCh38
Transcripts
NM_003648.2 NM_152879.2 XM_005246107.3 XM_005246108.1 XM_011512028.2 XM_011512029.2 XM_011512030.2 XM_011512031.2 XM_011512032.2 XM_011512033.2 XM_011512035.1 XM_011512037.2 XM_011512039.2 XM_011512040.2 XM_017005138.1 XM_017005139.1 XM_017005140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3838881 c.1081C>A p.L361I Chr.2: 233438375-233438375 on Build GRCh38
720203 c.1083C>A p.L361L Chr.2: 233438377-233438377 on Build GRCh38
6883097 c.1074C>T p.G358G Chr.2: 233438368-233438368 on Build GRCh38
1406357 c.1059C>G p.D353E Chr.2: 233438353-233438353 on Build GRCh38
7152092 c.887G>A p.R296K Chr.2: 233438313-233438313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773499405 Chr.2: 233438393-233438393 on Build GRCh38
rs920110767 Chr.2: 233438269-233438269 on Build GRCh38
rs59458664 Chr.2: 233438398-233438398 on Build GRCh38
rs370373545 Chr.2: 233438377-233438377 on Build GRCh38
rs3214826 Chr.2: 233438446-233438446 on Build GRCh38
rs369225332 Chr.2: 233438338-233438338 on Build GRCh38
rs763733034 Chr.2: 233438392-233438392 on Build GRCh38
rs755596405 Chr.2: 233438406-233438406 on Build GRCh38
rs770447761 Chr.2: 233438351-233438351 on Build GRCh38
rs377465249 Chr.2: 233438350-233438350 on Build GRCh38
rs756226321 Chr.2: 233438298-233438298 on Build GRCh38
rs760932836 Chr.2: 233438395-233438395 on Build GRCh38
rs775582380 Chr.2: 233438427-233438430 on Build GRCh38
rs370481507 Chr.2: 233438412-233438412 on Build GRCh38
rs186154470 Chr.2: 233438362-233438362 on Build GRCh38
rs1040313558 Chr.2: 233438433-233438433 on Build GRCh38
rs369752765 Chr.2: 233438322-233438322 on Build GRCh38
rs769697909 Chr.2: 233438358-233438358 on Build GRCh38
rs754424806 Chr.2: 233438304-233438304 on Build GRCh38
rs1047174948 Chr.2: 233438378-233438378 on Build GRCh38
rs376848918 Chr.2: 233438305-233438305 on Build GRCh38
rs148849509 Chr.2: 233438270-233438270 on Build GRCh38
rs373150668 Chr.2: 233438266-233438266 on Build GRCh38
rs147504312 Chr.2: 233438353-233438353 on Build GRCh38
rs765049780 Chr.2: 233438263-233438263 on Build GRCh38
rs750539012 Chr.2: 233438290-233438290 on Build GRCh38
rs892307506 Chr.2: 233438447-233438447 on Build GRCh38
rs753942821 Chr.2: 233438402-233438402 on Build GRCh38
rs758053252 Chr.2: 233438339-233438339 on Build GRCh38
rs767771309 Chr.2: 233438274-233438274 on Build GRCh38
rs758802900 Chr.2: 233438422-233438422 on Build GRCh38
rs768752099 Chr.2: 233438299-233438299 on Build GRCh38
rs765831491 Chr.2: 233438411-233438411 on Build GRCh38
rs777485542 Chr.2: 233438341-233438341 on Build GRCh38

View Full Product Details