Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99720202-99720473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A7
Entrez Gene ID
1551
Gene Name
cytochrome P450 family 3 subfamily A member 7
Gene Aliases
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
UniGene
Hs.111944
Gene Chromosome Location
Chr.7: 99705037-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
CYP3A7-CYP3A51P
Entrez Gene ID
100861540
Gene Name
CYP3A7-CYP3A51P readthrough
Gene Aliases
CYP3A7-3AP1, CYP3A7-CYP3AP1, CYP3A7.1L
UniGene
Gene Chromosome Location
Chr.7: 99684679-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
ZSCAN25
Entrez Gene ID
221785
Gene Name
zinc finger and SCAN domain containing 25
Gene Aliases
ZNF498
UniGene
Hs.446297
Gene Chromosome Location
Chr.7: 99616600-99738689 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3642935 c.315G>A p.R105R Chr.7: 99720316-99720316 on Build GRCh38
6111235 c.252C>T p.I84I Chr.7: 99720379-99720379 on Build GRCh38
7201205 c.274delA p.T92fs*3 Chr.7: 99720357-99720357 on Build GRCh38
453674 c.299C>T p.S100F Chr.7: 99720332-99720332 on Build GRCh38
1093874 c.262G>A p.D88N Chr.7: 99720369-99720369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750590665 Chr.7: 99720363-99720363 on Build GRCh38
rs917760217 Chr.7: 99720235-99720235 on Build GRCh38
rs754511331 Chr.7: 99720307-99720307 on Build GRCh38
rs200804987 Chr.7: 99720397-99720397 on Build GRCh38
rs748514156 Chr.7: 99720295-99720295 on Build GRCh38
rs781496507 Chr.7: 99720270-99720273 on Build GRCh38
rs751049219 Chr.7: 99720313-99720313 on Build GRCh38
rs374554025 Chr.7: 99720450-99720450 on Build GRCh38
rs200260866 Chr.7: 99720337-99720337 on Build GRCh38
rs778296509 Chr.7: 99720402-99720402 on Build GRCh38
rs781610168 Chr.7: 99720281-99720281 on Build GRCh38
rs774004460 Chr.7: 99720319-99720319 on Build GRCh38
rs545940540 Chr.7: 99720369-99720369 on Build GRCh38
rs763979975 Chr.7: 99720359-99720359 on Build GRCh38
rs1019283465 Chr.7: 99720354-99720354 on Build GRCh38
rs767410266 Chr.7: 99720331-99720331 on Build GRCh38
rs769460252 Chr.7: 99720304-99720304 on Build GRCh38
rs756386099 Chr.7: 99720395-99720395 on Build GRCh38
rs368291137 Chr.7: 99720277-99720280 on Build GRCh38
rs772014260 Chr.7: 99720441-99720441 on Build GRCh38
rs939876750 Chr.7: 99720256-99720256 on Build GRCh38
rs773982770 Chr.7: 99720422-99720422 on Build GRCh38
rs755491187 Chr.7: 99720376-99720376 on Build GRCh38
rs116248418 Chr.7: 99720318-99720318 on Build GRCh38
rs377456043 Chr.7: 99720429-99720429 on Build GRCh38
rs748724136 Chr.7: 99720383-99720383 on Build GRCh38
rs574715990 Chr.7: 99720317-99720317 on Build GRCh38
rs781593104 Chr.7: 99720381-99720381 on Build GRCh38
rs745416871 Chr.7: 99720433-99720433 on Build GRCh38
rs201450861 Chr.7: 99720329-99720329 on Build GRCh38
rs775331321 Chr.7: 99720335-99720335 on Build GRCh38
rs780126436 Chr.7: 99720268-99720268 on Build GRCh38
rs770776041 Chr.7: 99720411-99720411 on Build GRCh38
rs748884685 Chr.7: 99720406-99720406 on Build GRCh38
rs755442836 Chr.7: 99720277-99720277 on Build GRCh38
rs751698859 Chr.7: 99720373-99720373 on Build GRCh38
rs45466796 Chr.7: 99720370-99720370 on Build GRCh38
rs1036860532 Chr.7: 99720274-99720274 on Build GRCh38
rs144044787 Chr.7: 99720339-99720339 on Build GRCh38

View Full Product Details