Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179071838-179072111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20B
Entrez Gene ID
9917
Gene Name
family with sequence similarity 20 member B
Gene Aliases
gxk1
UniGene
Hs.5737
Gene Chromosome Location
Chr.1: 179025804-179076574 on Build GRCh38
Transcripts
NM_001324310.1 NM_001324311.1 NM_014864.3 XM_017003001.1 XM_017003002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659416 c.1119C>T p.D373D Chr.1: 179072033-179072033 on Build GRCh38
4682899 c.1120G>A p.A374T Chr.1: 179072034-179072034 on Build GRCh38
5616671 c.1008G>T p.V336V Chr.1: 179071922-179071922 on Build GRCh38
6659415 c.1004G>A p.R335Q Chr.1: 179071918-179071918 on Build GRCh38
6659404 c.1015T>C p.W339R Chr.1: 179071929-179071929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754374566 Chr.1: 179072010-179072010 on Build GRCh38
rs561162986 Chr.1: 179071862-179071862 on Build GRCh38
rs749604881 Chr.1: 179071920-179071920 on Build GRCh38
rs111556501 Chr.1: 179071896-179071896 on Build GRCh38
rs757259319 Chr.1: 179071897-179071897 on Build GRCh38
rs764204346 Chr.1: 179071882-179071882 on Build GRCh38
rs769610735 Chr.1: 179071983-179071983 on Build GRCh38
rs377519094 Chr.1: 179072034-179072034 on Build GRCh38
rs766740691 Chr.1: 179072009-179072009 on Build GRCh38
rs775114276 Chr.1: 179071872-179071872 on Build GRCh38
rs3820163 Chr.1: 179071876-179071876 on Build GRCh38
rs868540133 Chr.1: 179072051-179072051 on Build GRCh38
rs774110328 Chr.1: 179072089-179072089 on Build GRCh38
rs370601350 Chr.1: 179072069-179072069 on Build GRCh38
rs1035380528 Chr.1: 179071869-179071869 on Build GRCh38
rs754953535 Chr.1: 179071934-179071934 on Build GRCh38
rs556474499 Chr.1: 179071994-179071994 on Build GRCh38
rs374181238 Chr.1: 179072085-179072085 on Build GRCh38
rs200757870 Chr.1: 179072037-179072037 on Build GRCh38
rs112282462 Chr.1: 179072002-179072002 on Build GRCh38
rs202186782 Chr.1: 179072033-179072033 on Build GRCh38
rs201637462 Chr.1: 179071947-179071947 on Build GRCh38
rs777225732 Chr.1: 179071927-179071927 on Build GRCh38
rs746693808 Chr.1: 179071959-179071959 on Build GRCh38
rs200444001 Chr.1: 179071901-179071901 on Build GRCh38
rs767615051 Chr.1: 179071898-179071898 on Build GRCh38
rs146312839 Chr.1: 179071993-179071993 on Build GRCh38
rs144178097 Chr.1: 179072045-179072045 on Build GRCh38
rs755384540 Chr.1: 179071921-179071921 on Build GRCh38
rs201521365 Chr.1: 179071966-179071966 on Build GRCh38
rs145204794 Chr.1: 179071866-179071866 on Build GRCh38
rs765317086 Chr.1: 179071867-179071867 on Build GRCh38
rs762045477 Chr.1: 179072001-179072001 on Build GRCh38
rs200936773 Chr.1: 179071971-179071971 on Build GRCh38
rs756589376 Chr.1: 179071917-179071917 on Build GRCh38
rs780678801 Chr.1: 179071918-179071918 on Build GRCh38
rs775553811 Chr.1: 179071989-179071989 on Build GRCh38
rs755259109 Chr.1: 179072028-179072028 on Build GRCh38
rs760803150 Chr.1: 179072007-179072007 on Build GRCh38
rs745354379 Chr.1: 179072042-179072042 on Build GRCh38
rs146459916 Chr.1: 179072036-179072036 on Build GRCh38
rs779786849 Chr.1: 179072047-179072047 on Build GRCh38
rs746311048 Chr.1: 179071987-179071987 on Build GRCh38
rs554430970 Chr.1: 179071912-179071912 on Build GRCh38
rs750878446 Chr.1: 179071900-179071900 on Build GRCh38
rs887589979 Chr.1: 179071923-179071923 on Build GRCh38
rs144871480 Chr.1: 179072005-179072005 on Build GRCh38
rs774516389 Chr.1: 179072000-179072000 on Build GRCh38

View Full Product Details