Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 59833000-59833227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH3
Entrez Gene ID
81624
Gene Name
diaphanous related formin 3
Gene Aliases
AN, AUNA1, DIA2, DRF3, NSDAN, diap3, mDia2
UniGene
Hs.283127
Gene Chromosome Location
Chr.13: 59665583-60163985 on Build GRCh38
Transcripts
NM_001042517.1 NM_001258366.1 NM_001258367.1 NM_001258368.1 NM_001258369.1 NM_030932.3 XM_006719876.1 XM_011535258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275107 c.2918A>T p.N973I Chr.13: 59833216-59833216 on Build GRCh38
5494790 c.3014G>A p.R1005K Chr.13: 59833120-59833120 on Build GRCh38
1747489 c.3012C>G p.F1004L Chr.13: 59833122-59833122 on Build GRCh38
6286110 c.3017C>A p.T1006N Chr.13: 59833117-59833117 on Build GRCh38
7091759 c.3012C>T p.F1004F Chr.13: 59833122-59833122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749732852 Chr.13: 59833071-59833071 on Build GRCh38
rs866583748 Chr.13: 59833161-59833161 on Build GRCh38
rs775411792 Chr.13: 59833061-59833061 on Build GRCh38
rs751501612 Chr.13: 59833070-59833070 on Build GRCh38
rs188491955 Chr.13: 59833033-59833033 on Build GRCh38
rs749950199 Chr.13: 59833079-59833079 on Build GRCh38
rs759339741 Chr.13: 59833106-59833106 on Build GRCh38
rs1056118228 Chr.13: 59833063-59833063 on Build GRCh38
rs531346416 Chr.13: 59833163-59833163 on Build GRCh38
rs191777050 Chr.13: 59833096-59833096 on Build GRCh38
rs746019091 Chr.13: 59833147-59833147 on Build GRCh38
rs776786984 Chr.13: 59833038-59833038 on Build GRCh38
rs200914646 Chr.13: 59833160-59833160 on Build GRCh38
rs764090592 Chr.13: 59833065-59833065 on Build GRCh38
rs762889560 Chr.13: 59833064-59833064 on Build GRCh38
rs574768169 Chr.13: 59833028-59833028 on Build GRCh38
rs371248128 Chr.13: 59833158-59833158 on Build GRCh38
rs141626456 Chr.13: 59833036-59833036 on Build GRCh38
rs765571186 Chr.13: 59833179-59833179 on Build GRCh38
rs978946306 Chr.13: 59833110-59833110 on Build GRCh38
rs929083616 Chr.13: 59833128-59833128 on Build GRCh38
rs761586647 Chr.13: 59833165-59833165 on Build GRCh38
rs771476447 Chr.13: 59833114-59833114 on Build GRCh38
rs778047401 Chr.13: 59833194-59833194 on Build GRCh38
rs754642174 Chr.13: 59833189-59833189 on Build GRCh38
rs552878787 Chr.13: 59833092-59833093 on Build GRCh38
rs765563758 Chr.13: 59833060-59833060 on Build GRCh38
rs758416030 Chr.13: 59833162-59833162 on Build GRCh38
rs767552891 Chr.13: 59833176-59833176 on Build GRCh38
rs774297605 Chr.13: 59833164-59833164 on Build GRCh38
rs748485288 Chr.13: 59833100-59833100 on Build GRCh38
rs755714650 Chr.13: 59833083-59833083 on Build GRCh38
rs755693892 Chr.13: 59833182-59833182 on Build GRCh38
rs765909306 Chr.13: 59833185-59833185 on Build GRCh38
rs753441646 Chr.13: 59833186-59833186 on Build GRCh38
rs757237807 Chr.13: 59833077-59833077 on Build GRCh38
rs938985771 Chr.13: 59833039-59833039 on Build GRCh38
rs759612637 Chr.13: 59833056-59833056 on Build GRCh38
rs776569376 Chr.13: 59833116-59833116 on Build GRCh38
rs778135686 Chr.13: 59833101-59833101 on Build GRCh38
rs767162052 Chr.13: 59833150-59833150 on Build GRCh38
rs778902778 Chr.13: 59833093-59833093 on Build GRCh38
rs182679873 Chr.13: 59833140-59833140 on Build GRCh38
rs1005610578 Chr.13: 59833156-59833156 on Build GRCh38
rs745943798 Chr.13: 59833139-59833139 on Build GRCh38

View Full Product Details