Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46146125-46146392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTCD
Entrez Gene ID
10841
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
LCHC1
UniGene
Hs.415846
Gene Chromosome Location
Chr.21: 46135981-46156481 on Build GRCh38
Transcripts
NM_001320412.1 NM_006657.2 NM_206965.1 XM_006723961.3 XM_006723962.3 XM_011529434.2 XM_011529435.2 XM_011529436.2 XM_011529437.2 XM_011529439.2 XM_011529440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3551713 c.933C>T p.S311S Chr.21: 46146301-46146301 on Build GRCh38
1031563 c.926T>G p.L309R Chr.21: 46146308-46146308 on Build GRCh38
4875044 c.926T>G p.L309R Chr.21: 46146308-46146308 on Build GRCh38
1207628 c.958C>T p.R320W Chr.21: 46146276-46146276 on Build GRCh38
5802421 c.964_966delATC p.I322delI Chr.21: 46146268-46146270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907173603 Chr.21: 46146237-46146239 on Build GRCh38
rs768705350 Chr.21: 46146268-46146268 on Build GRCh38
rs374048282 Chr.21: 46146267-46146267 on Build GRCh38
rs1043013862 Chr.21: 46146204-46146204 on Build GRCh38
rs750187692 Chr.21: 46146346-46146346 on Build GRCh38
rs950620982 Chr.21: 46146318-46146318 on Build GRCh38
rs572833594 Chr.21: 46146207-46146207 on Build GRCh38
rs777087998 Chr.21: 46146312-46146312 on Build GRCh38
rs902812954 Chr.21: 46146205-46146205 on Build GRCh38
rs943103654 Chr.21: 46146152-46146152 on Build GRCh38
rs564843158 Chr.21: 46146214-46146214 on Build GRCh38
rs575835131 Chr.21: 46146165-46146165 on Build GRCh38
rs757166751 Chr.21: 46146317-46146317 on Build GRCh38
rs1000812210 Chr.21: 46146210-46146210 on Build GRCh38
rs751433805 Chr.21: 46146232-46146232 on Build GRCh38
rs780882900 Chr.21: 46146252-46146252 on Build GRCh38
rs778890059 Chr.21: 46146366-46146366 on Build GRCh38
rs80225292 Chr.21: 46146176-46146176 on Build GRCh38
rs908897070 Chr.21: 46146333-46146333 on Build GRCh38
rs141103624 Chr.21: 46146315-46146315 on Build GRCh38
rs745629384 Chr.21: 46146255-46146255 on Build GRCh38
rs192911499 Chr.21: 46146365-46146365 on Build GRCh38
rs1002751357 Chr.21: 46146284-46146284 on Build GRCh38
rs1028902060 Chr.21: 46146308-46146308 on Build GRCh38
rs774886831 Chr.21: 46146270-46146270 on Build GRCh38
rs1008854525 Chr.21: 46146337-46146337 on Build GRCh38
rs935850350 Chr.21: 46146182-46146182 on Build GRCh38
rs780309087 Chr.21: 46146358-46146358 on Build GRCh38
rs770013826 Chr.21: 46146263-46146263 on Build GRCh38
rs757981172 Chr.21: 46146227-46146227 on Build GRCh38
rs757795632 Chr.21: 46146341-46146342 on Build GRCh38
rs752384764 Chr.21: 46146301-46146301 on Build GRCh38
rs755896152 Chr.21: 46146354-46146354 on Build GRCh38
rs376165758 Chr.21: 46146228-46146228 on Build GRCh38
rs757119136 Chr.21: 46146248-46146248 on Build GRCh38
rs562640107 Chr.21: 46146302-46146302 on Build GRCh38
rs370540590 Chr.21: 46146237-46146237 on Build GRCh38
rs772464714 Chr.21: 46146370-46146370 on Build GRCh38
rs761036208 Chr.21: 46146289-46146289 on Build GRCh38
rs77740289 Chr.21: 46146275-46146275 on Build GRCh38
rs765051182 Chr.21: 46146292-46146292 on Build GRCh38
rs375883596 Chr.21: 46146273-46146273 on Build GRCh38
rs781124332 Chr.21: 46146326-46146326 on Build GRCh38
rs536442589 Chr.21: 46146172-46146172 on Build GRCh38
rs965786743 Chr.21: 46146158-46146158 on Build GRCh38
rs773469899 Chr.21: 46146286-46146286 on Build GRCh38
rs1054678991 Chr.21: 46146189-46146189 on Build GRCh38
rs150856432 Chr.21: 46146373-46146373 on Build GRCh38
rs747848507 Chr.21: 46146272-46146273 on Build GRCh38
rs1012762819 Chr.21: 46146229-46146229 on Build GRCh38
rs540247905 Chr.21: 46146213-46146213 on Build GRCh38
rs984417284 Chr.21: 46146329-46146329 on Build GRCh38
rs752262254 Chr.21: 46146220-46146220 on Build GRCh38
rs1039180255 Chr.21: 46146157-46146157 on Build GRCh38
rs933531660 Chr.21: 46146191-46146191 on Build GRCh38

View Full Product Details