Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182729657-182729896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
UniGene
Hs.516632
Gene Chromosome Location
Chr.2: 182716041-182780025 on Build GRCh38
Transcripts
NM_001271581.1 NM_018981.2 NR_073365.1 NR_073366.1 NR_073367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631645 c.650A>G p.H217R Chr.2: 182729864-182729864 on Build GRCh38
3990905 c.678G>T p.V226V Chr.2: 182729892-182729892 on Build GRCh38
7086303 c.652G>A p.G218R Chr.2: 182729866-182729866 on Build GRCh38
6248547 c.652G>T p.G218* Chr.2: 182729866-182729866 on Build GRCh38
5861292 c.662_664delCAA p.S221_K222>* Chr.2: 182729876-182729878 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971226734 Chr.2: 182729719-182729719 on Build GRCh38
rs771758358 Chr.2: 182729861-182729863 on Build GRCh38
rs79974409 Chr.2: 182729793-182729793 on Build GRCh38
rs765823613 Chr.2: 182729705-182729705 on Build GRCh38
rs771300522 Chr.2: 182729802-182729802 on Build GRCh38
rs1038331802 Chr.2: 182729803-182729803 on Build GRCh38
rs529278812 Chr.2: 182729764-182729764 on Build GRCh38
rs753157762 Chr.2: 182729829-182729829 on Build GRCh38
rs779355957 Chr.2: 182729818-182729818 on Build GRCh38
rs750240525 Chr.2: 182729865-182729865 on Build GRCh38
rs918619492 Chr.2: 182729813-182729813 on Build GRCh38
rs761835493 Chr.2: 182729853-182729853 on Build GRCh38
rs746160251 Chr.2: 182729822-182729822 on Build GRCh38
rs557761501 Chr.2: 182729777-182729778 on Build GRCh38
rs758346730 Chr.2: 182729802-182729807 on Build GRCh38
rs140936889 Chr.2: 182729857-182729857 on Build GRCh38
rs1016107357 Chr.2: 182729715-182729715 on Build GRCh38
rs971717898 Chr.2: 182729787-182729787 on Build GRCh38
rs374998022 Chr.2: 182729863-182729863 on Build GRCh38
rs113424839 Chr.2: 182729792-182729792 on Build GRCh38
rs980360242 Chr.2: 182729738-182729738 on Build GRCh38
rs930035618 Chr.2: 182729835-182729835 on Build GRCh38
rs772343316 Chr.2: 182729827-182729827 on Build GRCh38
rs768769076 Chr.2: 182729843-182729843 on Build GRCh38
rs563860965 Chr.2: 182729760-182729760 on Build GRCh38
rs530460047 Chr.2: 182729841-182729841 on Build GRCh38
rs544070464 Chr.2: 182729701-182729701 on Build GRCh38
rs776658635 Chr.2: 182729844-182729844 on Build GRCh38
rs963033891 Chr.2: 182729731-182729731 on Build GRCh38

View Full Product Details