Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94664934-94665165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP1
Entrez Gene ID
54845
Gene Name
epithelial splicing regulatory protein 1
Gene Aliases
RBM35A, RMB35A
UniGene
Hs.487471
Gene Chromosome Location
Chr.8: 94640999-94707466 on Build GRCh38
Transcripts
NM_001034915.2 NM_001122825.1 NM_001122826.1 NM_001122827.1 NM_017697.3 XM_005250991.3 XM_005250992.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4567639 c.799_800GG>AA p.G267K Chr.8: 94664970-94664971 on Build GRCh38
6656438 c.847C>A p.L283I Chr.8: 94665018-94665018 on Build GRCh38
5783723 c.826G>A p.E276K Chr.8: 94664997-94664997 on Build GRCh38
1102629 c.887A>T p.E296V Chr.8: 94665058-94665058 on Build GRCh38
6656436 c.847C>A p.L283I Chr.8: 94665018-94665018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368959736 Chr.8: 94664969-94664969 on Build GRCh38
rs772723160 Chr.8: 94665044-94665044 on Build GRCh38
rs376603249 Chr.8: 94665114-94665114 on Build GRCh38
rs747729420 Chr.8: 94665031-94665031 on Build GRCh38
rs758588745 Chr.8: 94665130-94665130 on Build GRCh38
rs141654162 Chr.8: 94665081-94665081 on Build GRCh38
rs993596589 Chr.8: 94665014-94665014 on Build GRCh38
rs746242699 Chr.8: 94665123-94665123 on Build GRCh38
rs552466771 Chr.8: 94665135-94665135 on Build GRCh38
rs746769438 Chr.8: 94665007-94665007 on Build GRCh38
rs533561075 Chr.8: 94665091-94665091 on Build GRCh38
rs769194906 Chr.8: 94665042-94665042 on Build GRCh38
rs372591846 Chr.8: 94665047-94665047 on Build GRCh38
rs747108794 Chr.8: 94665114-94665115 on Build GRCh38
rs781277695 Chr.8: 94665121-94665121 on Build GRCh38
rs754337193 Chr.8: 94665095-94665095 on Build GRCh38
rs777960348 Chr.8: 94665112-94665113 on Build GRCh38
rs35313474 Chr.8: 94665086-94665087 on Build GRCh38
rs553998616 Chr.8: 94665123-94665123 on Build GRCh38
rs768764209 Chr.8: 94665009-94665009 on Build GRCh38
rs779735850 Chr.8: 94664974-94664974 on Build GRCh38
rs764640211 Chr.8: 94665092-94665092 on Build GRCh38
rs765244651 Chr.8: 94665115-94665115 on Build GRCh38
rs781262951 Chr.8: 94665026-94665026 on Build GRCh38
rs771129422 Chr.8: 94665117-94665118 on Build GRCh38
rs775868293 Chr.8: 94665089-94665089 on Build GRCh38
rs750612777 Chr.8: 94664968-94664968 on Build GRCh38

View Full Product Details