Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53801546-53801771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERLEC1
Entrez Gene ID
27248
Gene Name
endoplasmic reticulum lectin 1
Gene Aliases
C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
UniGene
Hs.438336
Gene Chromosome Location
Chr.2: 53786931-53826928 on Build GRCh38
Transcripts
NM_001127397.2 NM_001127398.2 NM_015701.4 XM_011532766.1 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001127397.2 NM_001127398.2 NM_015701.4 XM_011532766.1 NM_001164165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69348 c.750-1G>A p.? Chr.2: 53801712-53801712 on Build GRCh38
7098817 c.689C>G p.T230R Chr.2: 53801560-53801560 on Build GRCh38
7305264 c.785A>G p.Q262R Chr.2: 53801748-53801748 on Build GRCh38
4834357 c.717G>A p.L239L Chr.2: 53801588-53801588 on Build GRCh38
6479071 c.737A>C p.H246P Chr.2: 53801608-53801608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772737069 Chr.2: 53801648-53801648 on Build GRCh38
rs544240244 Chr.2: 53801694-53801694 on Build GRCh38
rs994158203 Chr.2: 53801632-53801632 on Build GRCh38
rs747954309 Chr.2: 53801673-53801673 on Build GRCh38
rs756621853 Chr.2: 53801730-53801730 on Build GRCh38
rs140415318 Chr.2: 53801607-53801607 on Build GRCh38
rs771305488 Chr.2: 53801708-53801708 on Build GRCh38
rs200001770 Chr.2: 53801671-53801671 on Build GRCh38
rs780411050 Chr.2: 53801738-53801738 on Build GRCh38
rs771637849 Chr.2: 53801620-53801620 on Build GRCh38
rs759609747 Chr.2: 53801677-53801677 on Build GRCh38
rs201668452 Chr.2: 53801735-53801735 on Build GRCh38
rs763184193 Chr.2: 53801686-53801686 on Build GRCh38
rs775159236 Chr.2: 53801685-53801685 on Build GRCh38
rs555612927 Chr.2: 53801661-53801661 on Build GRCh38
rs34142625 Chr.2: 53801577-53801577 on Build GRCh38
rs764255409 Chr.2: 53801693-53801693 on Build GRCh38
rs575611975 Chr.2: 53801668-53801668 on Build GRCh38
rs758727969 Chr.2: 53801594-53801594 on Build GRCh38
rs765012132 Chr.2: 53801678-53801678 on Build GRCh38
rs776696511 Chr.2: 53801672-53801672 on Build GRCh38
rs1034439594 Chr.2: 53801726-53801726 on Build GRCh38
rs750929390 Chr.2: 53801720-53801720 on Build GRCh38
rs757306066 Chr.2: 53801695-53801695 on Build GRCh38
rs371108781 Chr.2: 53801657-53801657 on Build GRCh38
rs767641763 Chr.2: 53801698-53801698 on Build GRCh38

View Full Product Details