Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50256253-50256480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAI2
Entrez Gene ID
2771
Gene Name
G protein subunit alpha i2
Gene Aliases
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1
UniGene
Hs.77269
Gene Chromosome Location
Chr.3: 50226688-50259354 on Build GRCh38
Transcripts
NM_001166425.1 NM_001282617.1 NM_001282618.1 NM_001282619.1 NM_001282620.1 NM_002070.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4171250 c.494A>T p.K165M Chr.3: 50256269-50256269 on Build GRCh38
2851113 c.539T>C p.V180A Chr.3: 50256266-50256266 on Build GRCh38
2851112 c.535C>T p.R179C Chr.3: 50256262-50256262 on Build GRCh38
5129696 c.495G>A p.K165K Chr.3: 50256270-50256270 on Build GRCh38
4325265 c.491T>C p.V164A Chr.3: 50256266-50256266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372346600 Chr.3: 50256341-50256341 on Build GRCh38
rs984808431 Chr.3: 50256320-50256320 on Build GRCh38
rs149019479 Chr.3: 50256309-50256309 on Build GRCh38
rs961330271 Chr.3: 50256352-50256352 on Build GRCh38
rs149208832 Chr.3: 50256365-50256365 on Build GRCh38
rs781839068 Chr.3: 50256349-50256349 on Build GRCh38
rs782378562 Chr.3: 50256327-50256327 on Build GRCh38
rs935631141 Chr.3: 50256436-50256436 on Build GRCh38
rs892014888 Chr.3: 50256444-50256444 on Build GRCh38
rs951992375 Chr.3: 50256298-50256298 on Build GRCh38
rs376981591 Chr.3: 50256347-50256347 on Build GRCh38
rs1054147007 Chr.3: 50256438-50256438 on Build GRCh38
rs782034383 Chr.3: 50256338-50256338 on Build GRCh38
rs782082674 Chr.3: 50256355-50256355 on Build GRCh38
rs927053482 Chr.3: 50256430-50256430 on Build GRCh38
rs782367988 Chr.3: 50256339-50256339 on Build GRCh38
rs185731969 Chr.3: 50256377-50256377 on Build GRCh38
rs373273275 Chr.3: 50256371-50256371 on Build GRCh38
rs781955898 Chr.3: 50256282-50256282 on Build GRCh38
rs979857704 Chr.3: 50256420-50256420 on Build GRCh38

View Full Product Details