Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128222034-128222307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
UniGene
Hs.522413
Gene Chromosome Location
Chr.9: 128203355-128255248 on Build GRCh38
Transcripts
NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7121078 c.922G>C p.E308Q Chr.9: 128222269-128222269 on Build GRCh38
1733112 c.953G>A p.R318H Chr.9: 128222300-128222300 on Build GRCh38
5510376 c.857C>T p.T286M Chr.9: 128222204-128222204 on Build GRCh38
3375097 c.936G>A p.E312E Chr.9: 128222283-128222283 on Build GRCh38
5875603 c.890G>A p.R297Q Chr.9: 128222237-128222237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993356731 Chr.9: 128222100-128222100 on Build GRCh38
rs763158060 Chr.9: 128222179-128222179 on Build GRCh38
rs758110039 Chr.9: 128222220-128222220 on Build GRCh38
rs3003608 Chr.9: 128222151-128222151 on Build GRCh38
rs757034927 Chr.9: 128222235-128222235 on Build GRCh38
rs781007168 Chr.9: 128222245-128222245 on Build GRCh38
rs964177691 Chr.9: 128222113-128222113 on Build GRCh38
rs745752040 Chr.9: 128222264-128222264 on Build GRCh38
rs752460388 Chr.9: 128222205-128222205 on Build GRCh38
rs769831767 Chr.9: 128222283-128222283 on Build GRCh38
rs745593935 Chr.9: 128222150-128222156 on Build GRCh38
rs774680078 Chr.9: 128222182-128222182 on Build GRCh38
rs559572430 Chr.9: 128222229-128222229 on Build GRCh38
rs745958902 Chr.9: 128222156-128222156 on Build GRCh38
rs764505046 Chr.9: 128222180-128222180 on Build GRCh38
rs762350766 Chr.9: 128222185-128222185 on Build GRCh38
rs557093654 Chr.9: 128222158-128222158 on Build GRCh38
rs777759824 Chr.9: 128222221-128222221 on Build GRCh38
rs767943937 Chr.9: 128222187-128222187 on Build GRCh38
rs911338676 Chr.9: 128222105-128222105 on Build GRCh38
rs754456952 Chr.9: 128222191-128222191 on Build GRCh38
rs960196208 Chr.9: 128222065-128222065 on Build GRCh38
rs951478214 Chr.9: 128222151-128222151 on Build GRCh38
rs539621689 Chr.9: 128222193-128222193 on Build GRCh38
rs879760095 Chr.9: 128222088-128222088 on Build GRCh38
rs1052613908 Chr.9: 128222267-128222267 on Build GRCh38
rs750891102 Chr.9: 128222189-128222189 on Build GRCh38
rs546034278 Chr.9: 128222200-128222200 on Build GRCh38
rs933803259 Chr.9: 128222236-128222236 on Build GRCh38
rs879179710 Chr.9: 128222085-128222085 on Build GRCh38
rs182060568 Chr.9: 128222174-128222174 on Build GRCh38

View Full Product Details