Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66725781-66726010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1 XM_017023008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851181 c.1363A>T p.T455S Chr.16: 66725843-66725843 on Build GRCh38
4607649 c.1362C>A p.S454R Chr.16: 66725844-66725844 on Build GRCh38
4061683 c.1339C>T p.P447S Chr.16: 66725867-66725867 on Build GRCh38
703928 c.1330C>T p.P444S Chr.16: 66725876-66725876 on Build GRCh38
972197 c.1321A>G p.T441A Chr.16: 66725885-66725885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368490019 Chr.16: 66725960-66725960 on Build GRCh38
rs745827809 Chr.16: 66725979-66725979 on Build GRCh38
rs746190108 Chr.16: 66725949-66725949 on Build GRCh38
rs757233087 Chr.16: 66725964-66725964 on Build GRCh38
rs143707837 Chr.16: 66725900-66725900 on Build GRCh38
rs748971013 Chr.16: 66725858-66725858 on Build GRCh38
rs1015922794 Chr.16: 66725845-66725845 on Build GRCh38
rs199572341 Chr.16: 66725959-66725959 on Build GRCh38
rs760622533 Chr.16: 66725910-66725910 on Build GRCh38
rs781238558 Chr.16: 66725975-66725975 on Build GRCh38
rs755508824 Chr.16: 66725980-66725980 on Build GRCh38
rs753942143 Chr.16: 66725957-66725957 on Build GRCh38
rs762283465 Chr.16: 66725878-66725878 on Build GRCh38
rs768910291 Chr.16: 66725957-66725960 on Build GRCh38
rs768894802 Chr.16: 66725867-66725867 on Build GRCh38
rs17852314 Chr.16: 66725832-66725832 on Build GRCh38
rs750892737 Chr.16: 66725962-66725963 on Build GRCh38
rs559136731 Chr.16: 66725945-66725945 on Build GRCh38
rs139479968 Chr.16: 66725871-66725871 on Build GRCh38
rs768051013 Chr.16: 66725894-66725894 on Build GRCh38
rs972987241 Chr.16: 66725944-66725944 on Build GRCh38
rs779423180 Chr.16: 66725963-66725963 on Build GRCh38

View Full Product Details