Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108718654-108718879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC7
Entrez Gene ID
163479
Gene Name
fibronectin type III domain containing 7
Gene Aliases
-
UniGene
Hs.258253
Gene Chromosome Location
Chr.1: 108712934-108742749 on Build GRCh38
Transcripts
NM_001144937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5024558 c.415G>C p.A139P Chr.1: 108718866-108718866 on Build GRCh38
2116926 c.401A>G p.E134G Chr.1: 108718852-108718852 on Build GRCh38
5661758 c.398delG p.E134fs*32 Chr.1: 108718849-108718849 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023728993 Chr.1: 108718842-108718842 on Build GRCh38
rs569504951 Chr.1: 108718811-108718811 on Build GRCh38
rs536333224 Chr.1: 108718777-108718777 on Build GRCh38
rs969285791 Chr.1: 108718837-108718837 on Build GRCh38
rs142736840 Chr.1: 108718699-108718700 on Build GRCh38
rs765002649 Chr.1: 108718809-108718809 on Build GRCh38
rs774622669 Chr.1: 108718847-108718847 on Build GRCh38
rs1034830759 Chr.1: 108718849-108718849 on Build GRCh38
rs993882087 Chr.1: 108718701-108718701 on Build GRCh38
rs374050882 Chr.1: 108718778-108718778 on Build GRCh38
rs775760245 Chr.1: 108718798-108718800 on Build GRCh38
rs980241378 Chr.1: 108718843-108718843 on Build GRCh38
rs373403924 Chr.1: 108718770-108718770 on Build GRCh38

View Full Product Details