Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62824390-62824643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A3
Entrez Gene ID
1299
Gene Name
collagen type IX alpha 3
Gene Aliases
DJ885L7.4.1, EDM3, IDD, MED
UniGene
Hs.716639
Gene Chromosome Location
Chr.20: 62817062-62841159 on Build GRCh38
Transcripts
NM_001853.3 XM_011528545.1 XM_017027666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4499565 c.545C>T p.P182L Chr.20: 62824470-62824470 on Build GRCh38
127700 c.552G>T p.G184G Chr.20: 62824477-62824477 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766999523 Chr.20: 62824418-62824418 on Build GRCh38
rs756125092 Chr.20: 62824551-62824551 on Build GRCh38
rs750203808 Chr.20: 62824525-62824525 on Build GRCh38
rs760452163 Chr.20: 62824431-62824431 on Build GRCh38
rs758354138 Chr.20: 62824461-62824461 on Build GRCh38
rs200808104 Chr.20: 62824490-62824490 on Build GRCh38
rs750386417 Chr.20: 62824459-62824459 on Build GRCh38
rs45476191 Chr.20: 62824439-62824439 on Build GRCh38
rs45440599 Chr.20: 62824572-62824572 on Build GRCh38
rs771255280 Chr.20: 62824479-62824479 on Build GRCh38
rs886056909 Chr.20: 62824477-62824477 on Build GRCh38
rs374384361 Chr.20: 62824554-62824554 on Build GRCh38
rs138872362 Chr.20: 62824456-62824456 on Build GRCh38
rs373505033 Chr.20: 62824531-62824531 on Build GRCh38
rs150148851 Chr.20: 62824471-62824471 on Build GRCh38
rs530176764 Chr.20: 62824462-62824462 on Build GRCh38
rs761514701 Chr.20: 62824521-62824521 on Build GRCh38
rs563231938 Chr.20: 62824428-62824428 on Build GRCh38
rs760071674 Chr.20: 62824416-62824416 on Build GRCh38
rs776090061 Chr.20: 62824517-62824517 on Build GRCh38
rs775106866 Chr.20: 62824422-62824422 on Build GRCh38
rs138630583 Chr.20: 62824511-62824511 on Build GRCh38
rs374632922 Chr.20: 62824618-62824618 on Build GRCh38
rs765392378 Chr.20: 62824463-62824471 on Build GRCh38
rs967035697 Chr.20: 62824556-62824556 on Build GRCh38
rs925678632 Chr.20: 62824585-62824585 on Build GRCh38
rs748207382 Chr.20: 62824468-62824468 on Build GRCh38
rs925699667 Chr.20: 62824420-62824420 on Build GRCh38
rs765009694 Chr.20: 62824523-62824523 on Build GRCh38
rs777936617 Chr.20: 62824473-62824473 on Build GRCh38
rs2734514 Chr.20: 62824613-62824613 on Build GRCh38
rs372593417 Chr.20: 62824440-62824440 on Build GRCh38
rs567821554 Chr.20: 62824580-62824580 on Build GRCh38
rs749672875 Chr.20: 62824475-62824475 on Build GRCh38
rs377255816 Chr.20: 62824510-62824510 on Build GRCh38
rs905278346 Chr.20: 62824488-62824488 on Build GRCh38
rs942407447 Chr.20: 62824602-62824602 on Build GRCh38
rs1042772958 Chr.20: 62824407-62824407 on Build GRCh38
rs377658275 Chr.20: 62824534-62824534 on Build GRCh38
rs182505479 Chr.20: 62824560-62824560 on Build GRCh38
rs1022574790 Chr.20: 62824555-62824555 on Build GRCh38
rs1038990687 Chr.20: 62824594-62824594 on Build GRCh38
rs774770515 Chr.20: 62824503-62824503 on Build GRCh38
rs746399071 Chr.20: 62824508-62824508 on Build GRCh38
rs942459768 Chr.20: 62824426-62824426 on Build GRCh38
rs376077101 Chr.20: 62824578-62824579 on Build GRCh38
rs552427559 Chr.20: 62824539-62824539 on Build GRCh38
rs376280669 Chr.20: 62824581-62824581 on Build GRCh38
rs765234765 Chr.20: 62824458-62824458 on Build GRCh38
rs755325344 Chr.20: 62824464-62824465 on Build GRCh38
rs774658858 Chr.20: 62824411-62824411 on Build GRCh38
rs1010214960 Chr.20: 62824543-62824543 on Build GRCh38

View Full Product Details