Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7226222-7226481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL2
Entrez Gene ID
1856
Gene Name
dishevelled segment polarity protein 2
Gene Aliases
-
UniGene
Hs.118640
Gene Chromosome Location
Chr.17: 7225342-7234548 on Build GRCh38
Transcripts
NM_004422.2 XM_005256502.1 XM_017024298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2976645 c.1849G>A p.G617S Chr.17: 7226227-7226227 on Build GRCh38
4919333 c.1844A>C p.K615T Chr.17: 7226232-7226232 on Build GRCh38
271229 c.1761G>T p.E587D Chr.17: 7226422-7226422 on Build GRCh38
5187258 c.1811C>T p.T604M Chr.17: 7226265-7226265 on Build GRCh38
4769108 c.1724C>A p.T575N Chr.17: 7226459-7226459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760883380 Chr.17: 7226239-7226239 on Build GRCh38
rs748005728 Chr.17: 7226264-7226264 on Build GRCh38
rs371112172 Chr.17: 7226426-7226426 on Build GRCh38
rs745455027 Chr.17: 7226275-7226277 on Build GRCh38
rs762111480 Chr.17: 7226378-7226378 on Build GRCh38
rs372921737 Chr.17: 7226357-7226357 on Build GRCh38
rs766840095 Chr.17: 7226403-7226403 on Build GRCh38
rs370880170 Chr.17: 7226275-7226275 on Build GRCh38
rs757949522 Chr.17: 7226247-7226247 on Build GRCh38
rs747210791 Chr.17: 7226325-7226325 on Build GRCh38
rs773513204 Chr.17: 7226381-7226381 on Build GRCh38
rs752499366 Chr.17: 7226308-7226308 on Build GRCh38
rs367563224 Chr.17: 7226281-7226281 on Build GRCh38
rs752409376 Chr.17: 7226415-7226415 on Build GRCh38
rs775501377 Chr.17: 7226269-7226269 on Build GRCh38
rs764349752 Chr.17: 7226279-7226279 on Build GRCh38
rs755993922 Chr.17: 7226321-7226321 on Build GRCh38
rs941356299 Chr.17: 7226428-7226428 on Build GRCh38
rs377522537 Chr.17: 7226405-7226405 on Build GRCh38
rs1045544304 Chr.17: 7226259-7226259 on Build GRCh38
rs371737565 Chr.17: 7226330-7226330 on Build GRCh38
rs138493643 Chr.17: 7226427-7226427 on Build GRCh38
rs757188541 Chr.17: 7226444-7226444 on Build GRCh38
rs751086029 Chr.17: 7226304-7226304 on Build GRCh38
rs754263413 Chr.17: 7226241-7226241 on Build GRCh38
rs375432216 Chr.17: 7226346-7226346 on Build GRCh38
rs781121098 Chr.17: 7226262-7226262 on Build GRCh38
rs575155196 Chr.17: 7226240-7226240 on Build GRCh38
rs973148117 Chr.17: 7226290-7226290 on Build GRCh38
rs754651383 Chr.17: 7226307-7226307 on Build GRCh38
rs757743539 Chr.17: 7226242-7226242 on Build GRCh38
rs762306066 Chr.17: 7226289-7226289 on Build GRCh38
rs749895719 Chr.17: 7226244-7226244 on Build GRCh38
rs778747234 Chr.17: 7226451-7226451 on Build GRCh38
rs373873616 Chr.17: 7226358-7226358 on Build GRCh38
rs771633429 Chr.17: 7226275-7226275 on Build GRCh38
rs780086702 Chr.17: 7226276-7226276 on Build GRCh38
rs776949531 Chr.17: 7226280-7226280 on Build GRCh38
rs774027037 Chr.17: 7226274-7226275 on Build GRCh38
rs770514562 Chr.17: 7226248-7226249 on Build GRCh38
rs902996625 Chr.17: 7226370-7226371 on Build GRCh38
rs750956419 Chr.17: 7226287-7226287 on Build GRCh38
rs763500518 Chr.17: 7226392-7226392 on Build GRCh38
rs755903914 Chr.17: 7226421-7226421 on Build GRCh38
rs753626462 Chr.17: 7226431-7226431 on Build GRCh38
rs200484159 Chr.17: 7226349-7226349 on Build GRCh38
rs779516848 Chr.17: 7226248-7226248 on Build GRCh38
rs749586116 Chr.17: 7226255-7226255 on Build GRCh38
rs375513655 Chr.17: 7226380-7226380 on Build GRCh38
rs765905839 Chr.17: 7226295-7226295 on Build GRCh38
rs773310143 Chr.17: 7226265-7226265 on Build GRCh38
rs200099877 Chr.17: 7226407-7226407 on Build GRCh38
rs908006315 Chr.17: 7226268-7226268 on Build GRCh38
rs770832523 Chr.17: 7226333-7226333 on Build GRCh38

View Full Product Details