Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45097894-45098167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700437 c.3531G>T p.Q1177H Chr.15: 45098043-45098043 on Build GRCh38
6650195 c.3516-3delC p.? Chr.15: 45098061-45098061 on Build GRCh38
1373139 c.3537C>A p.F1179L Chr.15: 45098037-45098037 on Build GRCh38
1608415 c.3516-2A>G p.? Chr.15: 45098060-45098060 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201296941 Chr.15: 45097952-45097952 on Build GRCh38
rs757009915 Chr.15: 45097986-45097986 on Build GRCh38
rs781226985 Chr.15: 45098025-45098025 on Build GRCh38
rs553193882 Chr.15: 45098122-45098122 on Build GRCh38
rs762150396 Chr.15: 45098064-45098064 on Build GRCh38
rs374484023 Chr.15: 45097979-45097979 on Build GRCh38
rs759127457 Chr.15: 45098046-45098046 on Build GRCh38
rs140393165 Chr.15: 45098016-45098016 on Build GRCh38
rs745712188 Chr.15: 45098060-45098061 on Build GRCh38
rs566375398 Chr.15: 45097929-45097929 on Build GRCh38
rs758958089 Chr.15: 45097977-45097977 on Build GRCh38
rs778652423 Chr.15: 45098043-45098058 on Build GRCh38
rs775017655 Chr.15: 45098053-45098053 on Build GRCh38
rs754161282 Chr.15: 45097999-45097999 on Build GRCh38
rs963430775 Chr.15: 45098146-45098146 on Build GRCh38
rs770767928 Chr.15: 45098042-45098042 on Build GRCh38
rs774014641 Chr.15: 45097975-45097975 on Build GRCh38
rs754214422 Chr.15: 45098061-45098061 on Build GRCh38
rs929921319 Chr.15: 45097920-45097920 on Build GRCh38
rs751594966 Chr.15: 45098109-45098109 on Build GRCh38
rs753114754 Chr.15: 45097985-45097985 on Build GRCh38
rs138971379 Chr.15: 45098015-45098015 on Build GRCh38
rs375424670 Chr.15: 45098066-45098066 on Build GRCh38
rs758401090 Chr.15: 45098024-45098024 on Build GRCh38
rs749036312 Chr.15: 45098040-45098040 on Build GRCh38
rs537587602 Chr.15: 45098028-45098028 on Build GRCh38
rs1048422529 Chr.15: 45097925-45097925 on Build GRCh38
rs1030979502 Chr.15: 45098110-45098110 on Build GRCh38
rs771461888 Chr.15: 45098052-45098052 on Build GRCh38
rs148718959 Chr.15: 45098056-45098056 on Build GRCh38
rs756551273 Chr.15: 45097991-45097991 on Build GRCh38
rs909336628 Chr.15: 45098142-45098142 on Build GRCh38
rs953596695 Chr.15: 45098114-45098114 on Build GRCh38
rs369528536 Chr.15: 45098093-45098093 on Build GRCh38
rs1010149231 Chr.15: 45098077-45098077 on Build GRCh38
rs368395842 Chr.15: 45098058-45098058 on Build GRCh38
rs536459950 Chr.15: 45098076-45098076 on Build GRCh38
rs764517475 Chr.15: 45098059-45098059 on Build GRCh38
rs777719069 Chr.15: 45098035-45098035 on Build GRCh38
rs764464268 Chr.15: 45097993-45097993 on Build GRCh38
rs866582409 Chr.15: 45098021-45098021 on Build GRCh38
rs144254445 Chr.15: 45098043-45098043 on Build GRCh38
rs745999464 Chr.15: 45098034-45098034 on Build GRCh38
rs573137918 Chr.15: 45098135-45098135 on Build GRCh38
rs757473358 Chr.15: 45098000-45098000 on Build GRCh38

View Full Product Details