Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47082261-47082534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4Z1
Entrez Gene ID
199974
Gene Name
cytochrome P450 family 4 subfamily Z member 1
Gene Aliases
CYP4A20
UniGene
Hs.176588
Gene Chromosome Location
Chr.1: 47067488-47118320 on Build GRCh38
Transcripts
NM_178134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490466 c.389G>A p.G130D Chr.1: 47082358-47082358 on Build GRCh38
5715696 c.432C>G p.F144L Chr.1: 47082401-47082401 on Build GRCh38
2191580 c.482G>A p.R161Q Chr.1: 47082451-47082451 on Build GRCh38
2191577 c.367C>T p.R123* Chr.1: 47082336-47082336 on Build GRCh38
4769332 c.472G>A p.E158K Chr.1: 47082441-47082441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375381087 Chr.1: 47082324-47082324 on Build GRCh38
rs771066431 Chr.1: 47082374-47082378 on Build GRCh38
rs759732809 Chr.1: 47082466-47082468 on Build GRCh38
rs751469303 Chr.1: 47082401-47082401 on Build GRCh38
rs1502924 Chr.1: 47082300-47082300 on Build GRCh38
rs769199451 Chr.1: 47082333-47082333 on Build GRCh38
rs763768660 Chr.1: 47082483-47082483 on Build GRCh38
rs150940120 Chr.1: 47082432-47082432 on Build GRCh38
rs745994794 Chr.1: 47082306-47082306 on Build GRCh38
rs763928046 Chr.1: 47082393-47082393 on Build GRCh38
rs772732845 Chr.1: 47082339-47082339 on Build GRCh38
rs540867697 Chr.1: 47082357-47082357 on Build GRCh38
rs745363805 Chr.1: 47082453-47082453 on Build GRCh38
rs146576339 Chr.1: 47082378-47082378 on Build GRCh38
rs374204130 Chr.1: 47082476-47082476 on Build GRCh38
rs375669698 Chr.1: 47082450-47082450 on Build GRCh38
rs774973940 Chr.1: 47082336-47082336 on Build GRCh38
rs377426400 Chr.1: 47082287-47082287 on Build GRCh38
rs762445971 Chr.1: 47082337-47082337 on Build GRCh38
rs761614347 Chr.1: 47082496-47082496 on Build GRCh38
rs757146734 Chr.1: 47082403-47082403 on Build GRCh38
rs200076743 Chr.1: 47082441-47082441 on Build GRCh38
rs150189598 Chr.1: 47082448-47082448 on Build GRCh38
rs201511396 Chr.1: 47082340-47082340 on Build GRCh38
rs767293654 Chr.1: 47082505-47082505 on Build GRCh38
rs371139120 Chr.1: 47082466-47082466 on Build GRCh38
rs74073737 Chr.1: 47082297-47082297 on Build GRCh38
rs920975226 Chr.1: 47082368-47082368 on Build GRCh38
rs530391300 Chr.1: 47082470-47082470 on Build GRCh38
rs774087778 Chr.1: 47082488-47082488 on Build GRCh38
rs777684860 Chr.1: 47082288-47082288 on Build GRCh38
rs368708579 Chr.1: 47082451-47082451 on Build GRCh38
rs774483086 Chr.1: 47082377-47082377 on Build GRCh38
rs756224570 Chr.1: 47082431-47082431 on Build GRCh38
rs769863706 Chr.1: 47082310-47082310 on Build GRCh38
rs377639787 Chr.1: 47082359-47082359 on Build GRCh38
rs371107865 Chr.1: 47082366-47082366 on Build GRCh38
rs750416622 Chr.1: 47082405-47082405 on Build GRCh38
rs775955044 Chr.1: 47082349-47082349 on Build GRCh38
rs780339548 Chr.1: 47082319-47082319 on Build GRCh38
rs749569807 Chr.1: 47082439-47082439 on Build GRCh38
rs867712109 Chr.1: 47082347-47082347 on Build GRCh38
rs577678543 Chr.1: 47082404-47082404 on Build GRCh38
rs369935114 Chr.1: 47082294-47082294 on Build GRCh38

View Full Product Details