Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136942543-136942782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
UniGene
Hs.522507
Gene Chromosome Location
Chr.9: 136940433-136944774 on Build GRCh38
Transcripts
NM_018998.3 XM_005266089.2 XM_011518789.1 XM_017014812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691633 c.660C>G p.L220L Chr.9: 136942562-136942562 on Build GRCh38
3216751 c.608C>T p.S203L Chr.9: 136942614-136942614 on Build GRCh38
7367963 c.113C>T p.S38L Chr.9: 136942614-136942614 on Build GRCh38
4609834 c.573_574delGT p.F192fs*72 Chr.9: 136942648-136942649 on Build GRCh38
5599545 c.606C>T p.I202I Chr.9: 136942616-136942616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149656628 Chr.9: 136942574-136942574 on Build GRCh38
rs750768869 Chr.9: 136942699-136942700 on Build GRCh38
rs147650693 Chr.9: 136942599-136942599 on Build GRCh38
rs773949913 Chr.9: 136942569-136942569 on Build GRCh38
rs748635277 Chr.9: 136942640-136942640 on Build GRCh38
rs765657666 Chr.9: 136942684-136942684 on Build GRCh38
rs144989211 Chr.9: 136942725-136942726 on Build GRCh38
rs767788923 Chr.9: 136942581-136942581 on Build GRCh38
rs746089632 Chr.9: 136942742-136942742 on Build GRCh38
rs373825023 Chr.9: 136942753-136942753 on Build GRCh38
rs766954091 Chr.9: 136942757-136942757 on Build GRCh38
rs756243023 Chr.9: 136942626-136942626 on Build GRCh38
rs903693917 Chr.9: 136942667-136942667 on Build GRCh38
rs140196097 Chr.9: 136942674-136942674 on Build GRCh38
rs372187895 Chr.9: 136942709-136942709 on Build GRCh38
rs770421328 Chr.9: 136942651-136942651 on Build GRCh38
rs145441520 Chr.9: 136942577-136942577 on Build GRCh38
rs766079091 Chr.9: 136942761-136942761 on Build GRCh38
rs147681293 Chr.9: 136942668-136942668 on Build GRCh38
rs546645083 Chr.9: 136942737-136942737 on Build GRCh38
rs749869265 Chr.9: 136942759-136942759 on Build GRCh38
rs766087360 Chr.9: 136942709-136942710 on Build GRCh38
rs756434861 Chr.9: 136942703-136942704 on Build GRCh38
rs762840468 Chr.9: 136942670-136942670 on Build GRCh38
rs975725477 Chr.9: 136942718-136942718 on Build GRCh38
rs377379004 Chr.9: 136942594-136942594 on Build GRCh38
rs201523922 Chr.9: 136942719-136942719 on Build GRCh38
rs745710703 Chr.9: 136942664-136942664 on Build GRCh38
rs532707014 Chr.9: 136942688-136942688 on Build GRCh38
rs766782139 Chr.9: 136942628-136942628 on Build GRCh38
rs200351489 Chr.9: 136942596-136942596 on Build GRCh38
rs546064512 Chr.9: 136942717-136942717 on Build GRCh38
rs937921948 Chr.9: 136942669-136942669 on Build GRCh38
rs2292926 Chr.9: 136942619-136942619 on Build GRCh38
rs776227327 Chr.9: 136942601-136942601 on Build GRCh38
rs138718204 Chr.9: 136942685-136942685 on Build GRCh38
rs750690697 Chr.9: 136942622-136942622 on Build GRCh38
rs956441807 Chr.9: 136942578-136942578 on Build GRCh38
rs764563932 Chr.9: 136942725-136942725 on Build GRCh38
rs777365935 Chr.9: 136942643-136942643 on Build GRCh38
rs377326923 Chr.9: 136942756-136942756 on Build GRCh38
rs768515548 Chr.9: 136942702-136942702 on Build GRCh38
rs368747499 Chr.9: 136942708-136942708 on Build GRCh38
rs538857883 Chr.9: 136942754-136942754 on Build GRCh38
rs183917226 Chr.9: 136942652-136942652 on Build GRCh38
rs778366386 Chr.9: 136942737-136942738 on Build GRCh38
rs370546816 Chr.9: 136942725-136942731 on Build GRCh38
rs371335676 Chr.9: 136942672-136942672 on Build GRCh38
rs369275132 Chr.9: 136942595-136942595 on Build GRCh38
rs142133236 Chr.9: 136942600-136942600 on Build GRCh38
rs140995298 Chr.9: 136942673-136942673 on Build GRCh38
rs757328148 Chr.9: 136942580-136942580 on Build GRCh38
rs142701655 Chr.9: 136942714-136942715 on Build GRCh38
rs776839834 Chr.9: 136942562-136942562 on Build GRCh38
rs139868725 Chr.9: 136942567-136942567 on Build GRCh38
rs754536040 Chr.9: 136942736-136942737 on Build GRCh38
rs142670174 Chr.9: 136942597-136942597 on Build GRCh38
rs769612103 Chr.9: 136942697-136942697 on Build GRCh38
rs763403695 Chr.9: 136942724-136942724 on Build GRCh38
rs775396065 Chr.9: 136942613-136942613 on Build GRCh38
rs567366453 Chr.9: 136942760-136942760 on Build GRCh38
rs774342063 Chr.9: 136942706-136942706 on Build GRCh38
rs778755224 Chr.9: 136942733-136942733 on Build GRCh38
rs751766961 Chr.9: 136942681-136942681 on Build GRCh38
rs768021274 Chr.9: 136942635-136942641 on Build GRCh38
rs761571145 Chr.9: 136942572-136942572 on Build GRCh38
rs566278473 Chr.9: 136942610-136942610 on Build GRCh38
rs531880221 Chr.9: 136942726-136942726 on Build GRCh38
rs894643250 Chr.9: 136942625-136942625 on Build GRCh38
rs754085036 Chr.9: 136942630-136942630 on Build GRCh38
rs772984851 Chr.9: 136942573-136942573 on Build GRCh38
rs778462523 Chr.9: 136942675-136942675 on Build GRCh38
rs747657165 Chr.9: 136942754-136942755 on Build GRCh38
rs148935719 Chr.9: 136942686-136942686 on Build GRCh38
rs764242695 Chr.9: 136942576-136942576 on Build GRCh38
rs367779676 Chr.9: 136942736-136942736 on Build GRCh38
rs776659501 Chr.9: 136942575-136942575 on Build GRCh38
rs566880015 Chr.9: 136942729-136942729 on Build GRCh38
rs374271255 Chr.9: 136942700-136942700 on Build GRCh38
rs907454917 Chr.9: 136942648-136942648 on Build GRCh38
rs762193986 Chr.9: 136942587-136942588 on Build GRCh38
rs148415286 Chr.9: 136942590-136942590 on Build GRCh38
rs775839145 Chr.9: 136942748-136942748 on Build GRCh38
rs749150527 Chr.9: 136942730-136942730 on Build GRCh38
rs371142063 Chr.9: 136942732-136942732 on Build GRCh38
rs750625093 Chr.9: 136942587-136942587 on Build GRCh38
rs759782408 Chr.9: 136942710-136942710 on Build GRCh38
rs751094496 Chr.9: 136942687-136942687 on Build GRCh38
rs770203825 Chr.9: 136942743-136942743 on Build GRCh38
rs758012073 Chr.9: 136942641-136942641 on Build GRCh38
rs201987845 Chr.9: 136942631-136942631 on Build GRCh38
rs759520143 Chr.9: 136942605-136942605 on Build GRCh38
rs745504729 Chr.9: 136942694-136942694 on Build GRCh38
rs558779787 Chr.9: 136942645-136942645 on Build GRCh38
rs945128052 Chr.9: 136942636-136942636 on Build GRCh38
rs779779412 Chr.9: 136942698-136942698 on Build GRCh38
rs566561880 Chr.9: 136942739-136942739 on Build GRCh38
rs368508014 Chr.9: 136942727-136942727 on Build GRCh38
rs144433423 Chr.9: 136942614-136942614 on Build GRCh38

View Full Product Details