Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29282189-29282448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EWSR1
Entrez Gene ID
2130
Gene Name
EWS RNA binding protein 1
Gene Aliases
EWS, EWS-FLI1, bK984G1.4
UniGene
Hs.374477
Gene Chromosome Location
Chr.22: 29268009-29300525 on Build GRCh38
Transcripts
NM_001163285.1 NM_001163286.1 NM_001163287.1 NM_005243.3 NM_013986.3 XM_005261389.3 XM_005261390.4 XM_011529995.2 XM_011529996.2 XM_011529997.1 XM_011529998.1 XM_011529999.2 XM_011530000.1 XM_011530001.2 XM_011530002.2 XM_017028644.1 XM_017028645.1 XM_017028646.1 XM_017028647.1 XM_017028648.1 XM_017028649.1 XM_017028650.1 XM_017028651.1 XM_017028652.1 XM_017028653.1 XM_017028654.1 XM_017028655.1 XM_017028656.1 XM_017028657.1 XM_017028658.1 XM_017028659.1 XM_017028660.1 XM_017028661.1 XM_017028662.1 XM_017028663.1 XM_017028664.1 XM_017028665.1 XM_017028666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5530671 c.415C>T p.P139S Chr.22: 29282391-29282391 on Build GRCh38
5530669 c.433C>T p.P145S Chr.22: 29282391-29282391 on Build GRCh38
5530670 c.415C>T p.P139S Chr.22: 29282391-29282391 on Build GRCh38
6219972 c.448C>A p.Q150K Chr.22: 29282424-29282424 on Build GRCh38
6219971 c.448C>A p.Q150K Chr.22: 29282424-29282424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753893849 Chr.22: 29282382-29282382 on Build GRCh38
rs1047752318 Chr.22: 29282329-29282329 on Build GRCh38
rs981508815 Chr.22: 29282241-29282241 on Build GRCh38
rs545290762 Chr.22: 29282350-29282350 on Build GRCh38
rs937507403 Chr.22: 29282249-29282249 on Build GRCh38
rs1055936962 Chr.22: 29282277-29282277 on Build GRCh38
rs28405089 Chr.22: 29282320-29282320 on Build GRCh38
rs1012966374 Chr.22: 29282347-29282347 on Build GRCh38
rs963738215 Chr.22: 29282259-29282259 on Build GRCh38
rs766555115 Chr.22: 29282381-29282381 on Build GRCh38
rs1019340706 Chr.22: 29282339-29282339 on Build GRCh38
rs1003503441 Chr.22: 29282384-29282384 on Build GRCh38
rs765207539 Chr.22: 29282392-29282392 on Build GRCh38
rs1008415872 Chr.22: 29282336-29282336 on Build GRCh38
rs758898123 Chr.22: 29282393-29282393 on Build GRCh38
rs940800424 Chr.22: 29282240-29282240 on Build GRCh38
rs1041808043 Chr.22: 29282309-29282309 on Build GRCh38
rs113613489 Chr.22: 29282425-29282425 on Build GRCh38
rs975064657 Chr.22: 29282269-29282269 on Build GRCh38
rs528905120 Chr.22: 29282246-29282246 on Build GRCh38
rs1045770015 Chr.22: 29282375-29282375 on Build GRCh38
rs139337869 Chr.22: 29282270-29282270 on Build GRCh38
rs760990655 Chr.22: 29282369-29282369 on Build GRCh38
rs8143120 Chr.22: 29282288-29282288 on Build GRCh38
rs575677339 Chr.22: 29282215-29282215 on Build GRCh38
rs376496961 Chr.22: 29282296-29282296 on Build GRCh38
rs759608425 Chr.22: 29282383-29282383 on Build GRCh38
rs746848022 Chr.22: 29282237-29282237 on Build GRCh38
rs914981668 Chr.22: 29282244-29282247 on Build GRCh38
rs780276797 Chr.22: 29282295-29282295 on Build GRCh38
rs756866342 Chr.22: 29282426-29282426 on Build GRCh38
rs548930651 Chr.22: 29282330-29282330 on Build GRCh38
rs985237179 Chr.22: 29282231-29282231 on Build GRCh38
rs146418156 Chr.22: 29282411-29282411 on Build GRCh38
rs200637771 Chr.22: 29282405-29282405 on Build GRCh38
rs768612993 Chr.22: 29282344-29282344 on Build GRCh38

View Full Product Details