Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46958946-46959173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA2
Entrez Gene ID
10294
Gene Name
DnaJ heat shock protein family (Hsp40) member A2
Gene Aliases
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2
UniGene
Hs.368078
Gene Chromosome Location
Chr.16: 46955362-46973714 on Build GRCh38
Transcripts
NM_005880.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1709115 c.1013C>T p.P338L Chr.16: 46959037-46959037 on Build GRCh38
5937683 c.1012C>T p.P338S Chr.16: 46959038-46959038 on Build GRCh38
4856310 c.979G>C p.G327R Chr.16: 46959071-46959071 on Build GRCh38
3771949 c.953C>T p.P318L Chr.16: 46959097-46959097 on Build GRCh38
970802 c.925G>A p.V309I Chr.16: 46959125-46959125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148745889 Chr.16: 46959030-46959030 on Build GRCh38
rs762465852 Chr.16: 46959057-46959057 on Build GRCh38
rs775115873 Chr.16: 46959054-46959054 on Build GRCh38
rs773950249 Chr.16: 46959097-46959097 on Build GRCh38
rs377422570 Chr.16: 46959101-46959101 on Build GRCh38
rs1049292742 Chr.16: 46959073-46959073 on Build GRCh38
rs990469854 Chr.16: 46958978-46958978 on Build GRCh38
rs142308620 Chr.16: 46959129-46959129 on Build GRCh38
rs200198931 Chr.16: 46959032-46959032 on Build GRCh38
rs372523926 Chr.16: 46959096-46959096 on Build GRCh38
rs779094830 Chr.16: 46958993-46958993 on Build GRCh38
rs780937485 Chr.16: 46959026-46959026 on Build GRCh38
rs891852246 Chr.16: 46959042-46959042 on Build GRCh38
rs748554053 Chr.16: 46959010-46959010 on Build GRCh38
rs767723759 Chr.16: 46959134-46959137 on Build GRCh38
rs369080423 Chr.16: 46959062-46959062 on Build GRCh38
rs768387409 Chr.16: 46959094-46959094 on Build GRCh38
rs772512918 Chr.16: 46959019-46959019 on Build GRCh38
rs767478496 Chr.16: 46959127-46959127 on Build GRCh38
rs186339002 Chr.16: 46959039-46959039 on Build GRCh38

View Full Product Details