Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31428202-31428429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6A2
Entrez Gene ID
1339
Gene Name
cytochrome c oxidase subunit 6A2
Gene Aliases
COX6AH, COXVIAH
UniGene
Hs.250760
Gene Chromosome Location
Chr.16: 31427731-31428428 on Build GRCh38
Transcripts
NM_005205.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634351 c.41G>T p.S14I Chr.16: 31428285-31428285 on Build GRCh38
5817273 c.42C>T p.S14S Chr.16: 31428284-31428284 on Build GRCh38
7303828 c.73+2T>C p.? Chr.16: 31428251-31428251 on Build GRCh38
6334874 c.38C>A p.A13D Chr.16: 31428288-31428288 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767531048 Chr.16: 31428239-31428239 on Build GRCh38
rs765055668 Chr.16: 31428311-31428311 on Build GRCh38
rs892424165 Chr.16: 31428366-31428366 on Build GRCh38
rs778776784 Chr.16: 31428248-31428248 on Build GRCh38
rs753811433 Chr.16: 31428300-31428300 on Build GRCh38
rs745458434 Chr.16: 31428249-31428249 on Build GRCh38
rs771269329 Chr.16: 31428274-31428274 on Build GRCh38
rs753626288 Chr.16: 31428244-31428244 on Build GRCh38
rs909602856 Chr.16: 31428253-31428253 on Build GRCh38
rs760787848 Chr.16: 31428298-31428298 on Build GRCh38
rs890216794 Chr.16: 31428406-31428406 on Build GRCh38
rs763291425 Chr.16: 31428273-31428273 on Build GRCh38
rs763978974 Chr.16: 31428299-31428299 on Build GRCh38
rs115157197 Chr.16: 31428241-31428241 on Build GRCh38
rs200780049 Chr.16: 31428324-31428324 on Build GRCh38
rs752729509 Chr.16: 31428297-31428297 on Build GRCh38
rs73538329 Chr.16: 31428376-31428376 on Build GRCh38
rs779669707 Chr.16: 31428262-31428262 on Build GRCh38
rs200721543 Chr.16: 31428308-31428308 on Build GRCh38
rs748711535 Chr.16: 31428267-31428267 on Build GRCh38
rs370508400 Chr.16: 31428259-31428259 on Build GRCh38
rs140129800 Chr.16: 31428292-31428292 on Build GRCh38
rs779830938 Chr.16: 31428333-31428333 on Build GRCh38
rs750129888 Chr.16: 31428318-31428318 on Build GRCh38
rs941118349 Chr.16: 31428271-31428271 on Build GRCh38
rs757046966 Chr.16: 31428247-31428247 on Build GRCh38
rs1031463975 Chr.16: 31428281-31428281 on Build GRCh38
rs552611628 Chr.16: 31428286-31428286 on Build GRCh38
rs368585114 Chr.16: 31428361-31428361 on Build GRCh38
rs767619130 Chr.16: 31428296-31428296 on Build GRCh38
rs1051501357 Chr.16: 31428391-31428391 on Build GRCh38
rs200209655 Chr.16: 31428328-31428328 on Build GRCh38
rs1037995114 Chr.16: 31428354-31428354 on Build GRCh38
rs773734363 Chr.16: 31428270-31428270 on Build GRCh38
rs374752640 Chr.16: 31428370-31428370 on Build GRCh38
rs60662915 Chr.16: 31428227-31428227 on Build GRCh38
rs1002300977 Chr.16: 31428388-31428388 on Build GRCh38
rs956100607 Chr.16: 31428282-31428282 on Build GRCh38
rs367655225 Chr.16: 31428350-31428350 on Build GRCh38
rs749748326 Chr.16: 31428353-31428353 on Build GRCh38
rs752639598 Chr.16: 31428240-31428240 on Build GRCh38
rs759672607 Chr.16: 31428229-31428229 on Build GRCh38
rs754503300 Chr.16: 31428342-31428342 on Build GRCh38
rs772329947 Chr.16: 31428375-31428375 on Build GRCh38
rs988229644 Chr.16: 31428283-31428283 on Build GRCh38
rs376029051 Chr.16: 31428360-31428360 on Build GRCh38
rs746537698 Chr.16: 31428338-31428338 on Build GRCh38
rs770249533 Chr.16: 31428268-31428268 on Build GRCh38

View Full Product Details