Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37744478-37744731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERLIN2
Entrez Gene ID
11160
Gene Name
ER lipid raft associated 2
Gene Aliases
C8orf2, Erlin-2, NET32, SPFH2, SPG18
UniGene
Hs.705490
Gene Chromosome Location
Chr.8: 37736579-37757801 on Build GRCh38
Transcripts
NM_001003790.3 NM_001003791.2 NM_007175.6 XM_005273392.2 XM_006716280.2 XM_017013000.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1472120 c.304G>A p.D102N Chr.8: 37744576-37744576 on Build GRCh38
5409526 c.415G>A p.E139K Chr.8: 37744687-37744687 on Build GRCh38
3699064 c.395C>T p.T132M Chr.8: 37744667-37744667 on Build GRCh38
1177323 c.366C>T p.H122H Chr.8: 37744638-37744638 on Build GRCh38
6022300 c.353A>C p.N118T Chr.8: 37744625-37744625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762587770 Chr.8: 37744668-37744668 on Build GRCh38
rs4739541 Chr.8: 37744562-37744562 on Build GRCh38
rs925560594 Chr.8: 37744527-37744527 on Build GRCh38
rs770914507 Chr.8: 37744687-37744687 on Build GRCh38
rs772986050 Chr.8: 37744667-37744667 on Build GRCh38
rs752368171 Chr.8: 37744571-37744571 on Build GRCh38
rs747207286 Chr.8: 37744660-37744660 on Build GRCh38
rs778852699 Chr.8: 37744564-37744565 on Build GRCh38
rs77074649 Chr.8: 37744504-37744504 on Build GRCh38
rs754387519 Chr.8: 37744564-37744564 on Build GRCh38
rs752389058 Chr.8: 37744632-37744632 on Build GRCh38
rs200292614 Chr.8: 37744656-37744656 on Build GRCh38
rs745986559 Chr.8: 37744608-37744608 on Build GRCh38
rs16887018 Chr.8: 37744590-37744590 on Build GRCh38
rs767231549 Chr.8: 37744568-37744568 on Build GRCh38
rs948623331 Chr.8: 37744517-37744517 on Build GRCh38
rs996380168 Chr.8: 37744685-37744685 on Build GRCh38
rs540027626 Chr.8: 37744582-37744582 on Build GRCh38
rs1001386205 Chr.8: 37744617-37744617 on Build GRCh38
rs75043299 Chr.8: 37744505-37744505 on Build GRCh38
rs1052410160 Chr.8: 37744575-37744575 on Build GRCh38
rs765871378 Chr.8: 37744530-37744530 on Build GRCh38
rs549415141 Chr.8: 37744605-37744605 on Build GRCh38
rs372395378 Chr.8: 37744602-37744602 on Build GRCh38
rs887731732 Chr.8: 37744705-37744705 on Build GRCh38
rs371713211 Chr.8: 37744661-37744661 on Build GRCh38
rs756597769 Chr.8: 37744572-37744572 on Build GRCh38
rs201754176 Chr.8: 37744513-37744513 on Build GRCh38
rs560537773 Chr.8: 37744510-37744510 on Build GRCh38
rs374568375 Chr.8: 37744638-37744638 on Build GRCh38
rs751083873 Chr.8: 37744558-37744558 on Build GRCh38
rs774116062 Chr.8: 37744692-37744692 on Build GRCh38
rs868818363 Chr.8: 37744569-37744569 on Build GRCh38
rs1031026835 Chr.8: 37744626-37744626 on Build GRCh38

View Full Product Details