Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134917684-134917957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCN1
Entrez Gene ID
2219
Gene Name
ficolin 1
Gene Aliases
FCNM
UniGene
Hs.440898
Gene Chromosome Location
Chr.9: 134909585-134917960 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4408633 c.71A>G p.N24S Chr.9: 134917801-134917801 on Build GRCh38
7271105 c.90G>A p.A30A Chr.9: 134917782-134917782 on Build GRCh38
3779800 c.9G>C p.L3L Chr.9: 134917863-134917863 on Build GRCh38
1106626 c.36C>T p.L12L Chr.9: 134917836-134917836 on Build GRCh38
1718858 c.29_30GG>AA p.R10Q Chr.9: 134917842-134917843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866277450 Chr.9: 134917836-134917836 on Build GRCh38
rs1014523666 Chr.9: 134917729-134917729 on Build GRCh38
rs757908236 Chr.9: 134917867-134917867 on Build GRCh38
rs1023627909 Chr.9: 134917733-134917733 on Build GRCh38
rs1053111730 Chr.9: 134917706-134917706 on Build GRCh38
rs781494519 Chr.9: 134917890-134917890 on Build GRCh38
rs752106307 Chr.9: 134917882-134917882 on Build GRCh38
rs371385883 Chr.9: 134917758-134917758 on Build GRCh38
rs1055014647 Chr.9: 134917709-134917709 on Build GRCh38
rs753958057 Chr.9: 134917801-134917801 on Build GRCh38
rs531095841 Chr.9: 134917844-134917844 on Build GRCh38
rs565391562 Chr.9: 134917827-134917827 on Build GRCh38
rs141185154 Chr.9: 134917797-134917797 on Build GRCh38
rs541534890 Chr.9: 134917821-134917821 on Build GRCh38
rs756301950 Chr.9: 134917898-134917898 on Build GRCh38
rs936410941 Chr.9: 134917736-134917736 on Build GRCh38
rs139385211 Chr.9: 134917766-134917766 on Build GRCh38
rs56330318 Chr.9: 134917756-134917756 on Build GRCh38
rs746234917 Chr.9: 134917892-134917892 on Build GRCh38
rs764626950 Chr.9: 134917880-134917880 on Build GRCh38
rs150747379 Chr.9: 134917800-134917800 on Build GRCh38
rs759227452 Chr.9: 134917847-134917847 on Build GRCh38
rs978022104 Chr.9: 134917925-134917925 on Build GRCh38
rs144049283 Chr.9: 134917747-134917747 on Build GRCh38
rs773091298 Chr.9: 134917835-134917835 on Build GRCh38
rs369342575 Chr.9: 134917722-134917722 on Build GRCh38
rs769909709 Chr.9: 134917723-134917723 on Build GRCh38
rs748653376 Chr.9: 134917842-134917842 on Build GRCh38
rs917873130 Chr.9: 134917713-134917713 on Build GRCh38
rs895847498 Chr.9: 134917865-134917865 on Build GRCh38
rs377762723 Chr.9: 134917849-134917849 on Build GRCh38
rs746896194 Chr.9: 134917921-134917921 on Build GRCh38
rs780199778 Chr.9: 134917899-134917899 on Build GRCh38
rs773262942 Chr.9: 134917914-134917914 on Build GRCh38
rs902191511 Chr.9: 134917731-134917731 on Build GRCh38
rs747366532 Chr.9: 134917820-134917820 on Build GRCh38
rs866140313 Chr.9: 134917929-134917929 on Build GRCh38
rs1044692890 Chr.9: 134917889-134917889 on Build GRCh38
rs937845059 Chr.9: 134917707-134917707 on Build GRCh38
rs757780535 Chr.9: 134917885-134917885 on Build GRCh38
rs749383522 Chr.9: 134917911-134917911 on Build GRCh38
rs1027744049 Chr.9: 134917763-134917763 on Build GRCh38
rs1053586205 Chr.9: 134917855-134917855 on Build GRCh38
rs143189775 Chr.9: 134917783-134917783 on Build GRCh38
rs762956044 Chr.9: 134917831-134917831 on Build GRCh38
rs372958997 Chr.9: 134917843-134917843 on Build GRCh38
rs775572280 Chr.9: 134917829-134917829 on Build GRCh38
rs778197405 Chr.9: 134917806-134917806 on Build GRCh38
rs55712611 Chr.9: 134917935-134917935 on Build GRCh38
rs768659603 Chr.9: 134917912-134917912 on Build GRCh38
rs781367631 Chr.9: 134917822-134917822 on Build GRCh38
rs776902266 Chr.9: 134917721-134917721 on Build GRCh38
rs138985629 Chr.9: 134917832-134917832 on Build GRCh38
rs559839771 Chr.9: 134917823-134917823 on Build GRCh38
rs756412026 Chr.9: 134917782-134917782 on Build GRCh38
rs760605743 Chr.9: 134917772-134917772 on Build GRCh38
rs10858293 Chr.9: 134917839-134917839 on Build GRCh38
rs754256515 Chr.9: 134917720-134917720 on Build GRCh38

View Full Product Details