Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 40074930-40075199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAM
Entrez Gene ID
1826
Gene Name
DS cell adhesion molecule
Gene Aliases
CHD2, CHD2-42, CHD2-52
UniGene
Hs.397800
Gene Chromosome Location
Chr.21: 40010999-40847113 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 XM_017028281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4954002 c.4831G>T p.V1611L Chr.21: 40075094-40075094 on Build GRCh38
6649420 c.4839G>A p.L1613L Chr.21: 40075086-40075086 on Build GRCh38
4410966 c.4759delC p.L1587fs*1 Chr.21: 40075166-40075166 on Build GRCh38
725135 c.4726C>A p.L1576I Chr.21: 40075199-40075199 on Build GRCh38
7118002 c.4878G>T p.K1626N Chr.21: 40075047-40075047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779560899 Chr.21: 40075066-40075066 on Build GRCh38
rs765995928 Chr.21: 40075041-40075041 on Build GRCh38
rs201077680 Chr.21: 40075067-40075067 on Build GRCh38
rs773110897 Chr.21: 40075149-40075149 on Build GRCh38
rs780232729 Chr.21: 40075077-40075077 on Build GRCh38
rs768804179 Chr.21: 40075007-40075007 on Build GRCh38
rs751242984 Chr.21: 40075050-40075050 on Build GRCh38
rs371238790 Chr.21: 40075098-40075098 on Build GRCh38
rs780235810 Chr.21: 40075029-40075029 on Build GRCh38
rs759236431 Chr.21: 40075157-40075157 on Build GRCh38
rs780754561 Chr.21: 40075058-40075058 on Build GRCh38
rs765143978 Chr.21: 40075012-40075012 on Build GRCh38
rs7275294 Chr.21: 40075009-40075009 on Build GRCh38
rs1006167724 Chr.21: 40074953-40074953 on Build GRCh38
rs983357490 Chr.21: 40074979-40074979 on Build GRCh38
rs761875826 Chr.21: 40075118-40075118 on Build GRCh38
rs373840594 Chr.21: 40075021-40075021 on Build GRCh38
rs746223683 Chr.21: 40074999-40074999 on Build GRCh38
rs754542814 Chr.21: 40075052-40075052 on Build GRCh38
rs757899407 Chr.21: 40075064-40075064 on Build GRCh38
rs145500496 Chr.21: 40075162-40075162 on Build GRCh38
rs768696185 Chr.21: 40075101-40075101 on Build GRCh38
rs762705210 Chr.21: 40075026-40075026 on Build GRCh38
rs565114141 Chr.21: 40075109-40075109 on Build GRCh38
rs1015711198 Chr.21: 40074982-40074982 on Build GRCh38
rs750993445 Chr.21: 40075168-40075168 on Build GRCh38
rs913740674 Chr.21: 40075075-40075075 on Build GRCh38
rs772252904 Chr.21: 40075001-40075001 on Build GRCh38
rs367647810 Chr.21: 40075063-40075063 on Build GRCh38
rs7275460 Chr.21: 40075131-40075131 on Build GRCh38
rs772464092 Chr.21: 40075076-40075076 on Build GRCh38
rs945157466 Chr.21: 40075089-40075089 on Build GRCh38
rs534328405 Chr.21: 40075025-40075025 on Build GRCh38
rs1042694601 Chr.21: 40075091-40075091 on Build GRCh38
rs368391292 Chr.21: 40075155-40075155 on Build GRCh38
rs989761654 Chr.21: 40075074-40075074 on Build GRCh38
rs760717159 Chr.21: 40075006-40075006 on Build GRCh38
rs751243455 Chr.21: 40075156-40075156 on Build GRCh38
rs767263741 Chr.21: 40075159-40075159 on Build GRCh38
rs747461437 Chr.21: 40075018-40075018 on Build GRCh38
rs556682802 Chr.21: 40075154-40075154 on Build GRCh38
rs200109772 Chr.21: 40074987-40074987 on Build GRCh38
rs779431691 Chr.21: 40074994-40074994 on Build GRCh38
rs868349320 Chr.21: 40074955-40074955 on Build GRCh38
rs529976538 Chr.21: 40075161-40075161 on Build GRCh38
rs775754153 Chr.21: 40075003-40075003 on Build GRCh38

View Full Product Details