Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4446819-4447090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
UniGene
Hs.459779
Gene Chromosome Location
Chr.16: 4425805-4456775 on Build GRCh38
Transcripts
NM_001135110.2 NM_001286516.1 NM_005147.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970703 c.1045G>A p.D349N Chr.16: 4446934-4446934 on Build GRCh38
5646268 c.241G>A p.E81K Chr.16: 4446906-4446906 on Build GRCh38
4678644 c.138C>T p.H46H Chr.16: 4447009-4447009 on Build GRCh38
5850919 c.1027G>A p.G343S Chr.16: 4446916-4446916 on Build GRCh38
4678643 c.1120G>A p.V374M Chr.16: 4447009-4447009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745336716 Chr.16: 4447051-4447051 on Build GRCh38
rs762102563 Chr.16: 4447007-4447007 on Build GRCh38
rs200077748 Chr.16: 4446881-4446881 on Build GRCh38
rs767912888 Chr.16: 4447025-4447025 on Build GRCh38
rs746711710 Chr.16: 4447001-4447001 on Build GRCh38
rs752977018 Chr.16: 4446854-4446854 on Build GRCh38
rs758332226 Chr.16: 4447054-4447054 on Build GRCh38
rs747874869 Chr.16: 4446918-4446918 on Build GRCh38
rs1016433964 Chr.16: 4446909-4446909 on Build GRCh38
rs753807980 Chr.16: 4446873-4446873 on Build GRCh38
rs147789110 Chr.16: 4447062-4447062 on Build GRCh38
rs767298254 Chr.16: 4446843-4446843 on Build GRCh38
rs2304633 Chr.16: 4446972-4446972 on Build GRCh38
rs139563269 Chr.16: 4446994-4446994 on Build GRCh38
rs377444073 Chr.16: 4447017-4447017 on Build GRCh38
rs776772813 Chr.16: 4446958-4446958 on Build GRCh38
rs757792003 Chr.16: 4446889-4446889 on Build GRCh38
rs143480833 Chr.16: 4447013-4447013 on Build GRCh38
rs746949528 Chr.16: 4447060-4447060 on Build GRCh38
rs147345131 Chr.16: 4446993-4446993 on Build GRCh38
rs755806162 Chr.16: 4446982-4446982 on Build GRCh38
rs775365702 Chr.16: 4446997-4446997 on Build GRCh38
rs771906855 Chr.16: 4446936-4446936 on Build GRCh38
rs768491254 Chr.16: 4447002-4447002 on Build GRCh38
rs200303451 Chr.16: 4446927-4446927 on Build GRCh38
rs368716357 Chr.16: 4446933-4446933 on Build GRCh38
rs201017600 Chr.16: 4446892-4446892 on Build GRCh38
rs765447203 Chr.16: 4447009-4447009 on Build GRCh38
rs371363899 Chr.16: 4446865-4446865 on Build GRCh38
rs139593728 Chr.16: 4446915-4446915 on Build GRCh38
rs745793971 Chr.16: 4446934-4446934 on Build GRCh38
rs766881681 Chr.16: 4446976-4446976 on Build GRCh38
rs150596934 Chr.16: 4446908-4446908 on Build GRCh38
rs761862631 Chr.16: 4446969-4446969 on Build GRCh38
rs764160978 Chr.16: 4446953-4446953 on Build GRCh38
rs760789344 Chr.16: 4446868-4446868 on Build GRCh38
rs753484983 Chr.16: 4447033-4447033 on Build GRCh38
rs1020655374 Chr.16: 4446847-4446847 on Build GRCh38
rs776773756 Chr.16: 4447006-4447006 on Build GRCh38
rs1002516429 Chr.16: 4446859-4446859 on Build GRCh38
rs777225940 Chr.16: 4446986-4446986 on Build GRCh38
rs947237012 Chr.16: 4447023-4447023 on Build GRCh38
rs763575812 Chr.16: 4447014-4447014 on Build GRCh38
rs777365452 Chr.16: 4446921-4446921 on Build GRCh38
rs760993261 Chr.16: 4446949-4446949 on Build GRCh38
rs925548337 Chr.16: 4446906-4446906 on Build GRCh38
rs138637241 Chr.16: 4446943-4446943 on Build GRCh38
rs1041533892 Chr.16: 4447043-4447043 on Build GRCh38
rs1002715730 Chr.16: 4447061-4447061 on Build GRCh38
rs538999097 Chr.16: 4446850-4446850 on Build GRCh38
rs767982914 Chr.16: 4446867-4446868 on Build GRCh38
rs770860539 Chr.16: 4446930-4446930 on Build GRCh38
rs373301522 Chr.16: 4446919-4446919 on Build GRCh38
rs552548240 Chr.16: 4446864-4446864 on Build GRCh38
rs758942282 Chr.16: 4446974-4446974 on Build GRCh38
rs370464119 Chr.16: 4447050-4447050 on Build GRCh38
rs771962066 Chr.16: 4446996-4446996 on Build GRCh38
rs751822507 Chr.16: 4446980-4446980 on Build GRCh38
rs910528047 Chr.16: 4446983-4446983 on Build GRCh38
rs891096639 Chr.16: 4446932-4446932 on Build GRCh38
rs751958918 Chr.16: 4447018-4447018 on Build GRCh38
rs756874933 Chr.16: 4447041-4447041 on Build GRCh38
rs748967316 Chr.16: 4446990-4446990 on Build GRCh38
rs369923574 Chr.16: 4446907-4446907 on Build GRCh38
rs755367153 Chr.16: 4447021-4447021 on Build GRCh38
rs779919707 Chr.16: 4447056-4447056 on Build GRCh38
rs374888222 Chr.16: 4446872-4446872 on Build GRCh38
rs749431576 Chr.16: 4446869-4446869 on Build GRCh38

View Full Product Details