Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66482116-66482359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP3
Entrez Gene ID
10072
Gene Name
dipeptidyl peptidase 3
Gene Aliases
DPPIII
UniGene
Hs.502914
Gene Chromosome Location
Chr.11: 66480013-66509659 on Build GRCh38
Transcripts
NM_001256670.1 NM_005700.4 NM_130443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035731 c.113C>T p.S38F Chr.11: 66482313-66482313 on Build GRCh38
5565908 c.59G>A p.R20H Chr.11: 66482259-66482259 on Build GRCh38
1356220 c.98A>G p.Y33C Chr.11: 66482298-66482298 on Build GRCh38
338453 c.39C>T p.G13G Chr.11: 66482239-66482239 on Build GRCh38
5480328 c.71G>A p.R24H Chr.11: 66482271-66482271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748889499 Chr.11: 66482325-66482325 on Build GRCh38
rs139875519 Chr.11: 66482206-66482206 on Build GRCh38
rs746775000 Chr.11: 66482292-66482292 on Build GRCh38
rs562747542 Chr.11: 66482156-66482156 on Build GRCh38
rs780975451 Chr.11: 66482315-66482315 on Build GRCh38
rs756979162 Chr.11: 66482298-66482298 on Build GRCh38
rs201240945 Chr.11: 66482144-66482144 on Build GRCh38
rs955266815 Chr.11: 66482237-66482237 on Build GRCh38
rs147958933 Chr.11: 66482320-66482320 on Build GRCh38
rs200777816 Chr.11: 66482229-66482229 on Build GRCh38
rs777822910 Chr.11: 66482290-66482290 on Build GRCh38
rs900201852 Chr.11: 66482247-66482247 on Build GRCh38
rs747987185 Chr.11: 66482219-66482219 on Build GRCh38
rs200239534 Chr.11: 66482172-66482172 on Build GRCh38
rs145366083 Chr.11: 66482239-66482239 on Build GRCh38
rs199628557 Chr.11: 66482236-66482236 on Build GRCh38
rs756428183 Chr.11: 66482271-66482271 on Build GRCh38
rs557697442 Chr.11: 66482316-66482316 on Build GRCh38
rs201658473 Chr.11: 66482270-66482270 on Build GRCh38
rs753992246 Chr.11: 66482279-66482279 on Build GRCh38
rs769793568 Chr.11: 66482282-66482282 on Build GRCh38
rs199921998 Chr.11: 66482280-66482280 on Build GRCh38
rs200657045 Chr.11: 66482258-66482258 on Build GRCh38
rs763327052 Chr.11: 66482254-66482254 on Build GRCh38
rs75447316 Chr.11: 66482157-66482157 on Build GRCh38
rs879274099 Chr.11: 66482242-66482242 on Build GRCh38
rs1027625928 Chr.11: 66482330-66482330 on Build GRCh38
rs200857844 Chr.11: 66482283-66482283 on Build GRCh38
rs145932032 Chr.11: 66482205-66482205 on Build GRCh38
rs200192681 Chr.11: 66482244-66482244 on Build GRCh38
rs766527382 Chr.11: 66482273-66482273 on Build GRCh38
rs139018330 Chr.11: 66482259-66482259 on Build GRCh38
rs201828080 Chr.11: 66482240-66482240 on Build GRCh38
rs774828528 Chr.11: 66482321-66482321 on Build GRCh38
rs762427559 Chr.11: 66482333-66482333 on Build GRCh38
rs201562215 Chr.11: 66482175-66482175 on Build GRCh38
rs750530320 Chr.11: 66482181-66482181 on Build GRCh38
rs774858480 Chr.11: 66482331-66482331 on Build GRCh38
rs199998254 Chr.11: 66482264-66482264 on Build GRCh38
rs952257444 Chr.11: 66482190-66482190 on Build GRCh38
rs200614411 Chr.11: 66482329-66482329 on Build GRCh38
rs142478050 Chr.11: 66482265-66482265 on Build GRCh38
rs149689390 Chr.11: 66482234-66482234 on Build GRCh38
rs775414695 Chr.11: 66482305-66482305 on Build GRCh38

View Full Product Details