Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6623237-6623464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2040768 c.8309G>A p.R2770Q Chr.11: 6623367-6623367 on Build GRCh38
144658 c.8431C>T p.P2811S Chr.11: 6623245-6623245 on Build GRCh38
689872 c.8381C>T p.S2794L Chr.11: 6623295-6623295 on Build GRCh38
6348243 c.8302C>A p.R2768S Chr.11: 6623374-6623374 on Build GRCh38
5897783 c.8296G>T p.E2766* Chr.11: 6623380-6623380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778615533 Chr.11: 6623361-6623361 on Build GRCh38
rs757858922 Chr.11: 6623439-6623439 on Build GRCh38
rs939592673 Chr.11: 6623298-6623298 on Build GRCh38
rs776368310 Chr.11: 6623373-6623373 on Build GRCh38
rs748609148 Chr.11: 6623277-6623277 on Build GRCh38
rs773960003 Chr.11: 6623299-6623299 on Build GRCh38
rs72911011 Chr.11: 6623369-6623369 on Build GRCh38
rs781555020 Chr.11: 6623354-6623354 on Build GRCh38
rs770571515 Chr.11: 6623282-6623282 on Build GRCh38
rs868350000 Chr.11: 6623391-6623391 on Build GRCh38
rs200795986 Chr.11: 6623413-6623413 on Build GRCh38
rs200985379 Chr.11: 6623331-6623331 on Build GRCh38
rs374270843 Chr.11: 6623360-6623360 on Build GRCh38
rs751738610 Chr.11: 6623398-6623398 on Build GRCh38
rs775394503 Chr.11: 6623306-6623306 on Build GRCh38
rs773350586 Chr.11: 6623380-6623380 on Build GRCh38
rs764148393 Chr.11: 6623319-6623319 on Build GRCh38
rs200122562 Chr.11: 6623370-6623370 on Build GRCh38
rs760381368 Chr.11: 6623314-6623314 on Build GRCh38
rs763012228 Chr.11: 6623384-6623384 on Build GRCh38
rs999967170 Chr.11: 6623367-6623367 on Build GRCh38
rs372127346 Chr.11: 6623435-6623435 on Build GRCh38
rs375741388 Chr.11: 6623345-6623345 on Build GRCh38
rs758664651 Chr.11: 6623341-6623341 on Build GRCh38
rs951256878 Chr.11: 6623415-6623415 on Build GRCh38
rs373867079 Chr.11: 6623445-6623445 on Build GRCh38
rs368211314 Chr.11: 6623374-6623374 on Build GRCh38
rs771601009 Chr.11: 6623301-6623301 on Build GRCh38
rs778420931 Chr.11: 6623429-6623429 on Build GRCh38
rs748698718 Chr.11: 6623357-6623357 on Build GRCh38
rs200844364 Chr.11: 6623372-6623372 on Build GRCh38
rs761669414 Chr.11: 6623329-6623329 on Build GRCh38
rs141243126 Chr.11: 6623412-6623412 on Build GRCh38
rs373478460 Chr.11: 6623407-6623407 on Build GRCh38
rs750449956 Chr.11: 6623332-6623332 on Build GRCh38
rs747308612 Chr.11: 6623270-6623270 on Build GRCh38
rs753729735 Chr.11: 6623323-6623323 on Build GRCh38
rs752021858 Chr.11: 6623352-6623352 on Build GRCh38
rs765133574 Chr.11: 6623377-6623377 on Build GRCh38
rs769276323 Chr.11: 6623271-6623271 on Build GRCh38
rs138806709 Chr.11: 6623272-6623272 on Build GRCh38
rs745339829 Chr.11: 6623362-6623362 on Build GRCh38
rs766426409 Chr.11: 6623388-6623388 on Build GRCh38
rs755314374 Chr.11: 6623353-6623353 on Build GRCh38
rs771824096 Chr.11: 6623364-6623364 on Build GRCh38
rs756725661 Chr.11: 6623421-6623421 on Build GRCh38

View Full Product Details