Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 3567425-3567650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
UniGene
Hs.594640
Gene Chromosome Location
Chr.18: 3496032-4455307 on Build GRCh38
Transcripts
NM_001003809.2 NM_001242761.1 NM_001242762.1 NM_001242763.1 NM_001242764.1 NM_001242765.1 NM_001242766.1 NM_001308390.1 NM_004746.3 XM_005258171.2 XM_005258172.1 XM_005258173.3 XM_005258174.4 XM_006722367.3 XM_017026080.1 XM_017026081.1 XM_017026082.1 XM_017026083.1 XM_017026084.1 XM_017026085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364554 c.1088A>G p.D363G Chr.18: 3567577-3567577 on Build GRCh38
7157583 c.1085G>A p.G362E Chr.18: 3567514-3567514 on Build GRCh38
4275176 c.2013C>T p.P671P Chr.18: 3567534-3567534 on Build GRCh38
4866056 c.1146C>A p.S382S Chr.18: 3567519-3567519 on Build GRCh38
2811739 c.1975G>T p.A659S Chr.18: 3567572-3567572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202187623 Chr.18: 3567620-3567620 on Build GRCh38
rs373939141 Chr.18: 3567471-3567471 on Build GRCh38
rs752272792 Chr.18: 3567479-3567479 on Build GRCh38
rs745731417 Chr.18: 3567537-3567537 on Build GRCh38
rs61732678 Chr.18: 3567570-3567570 on Build GRCh38
rs1016127749 Chr.18: 3567477-3567477 on Build GRCh38
rs760944676 Chr.18: 3567473-3567473 on Build GRCh38
rs757947776 Chr.18: 3567481-3567481 on Build GRCh38
rs923218665 Chr.18: 3567591-3567591 on Build GRCh38
rs368372729 Chr.18: 3567558-3567558 on Build GRCh38
rs769452424 Chr.18: 3567548-3567548 on Build GRCh38
rs777229428 Chr.18: 3567486-3567486 on Build GRCh38
rs773582276 Chr.18: 3567615-3567615 on Build GRCh38
rs757116022 Chr.18: 3567518-3567518 on Build GRCh38
rs534513774 Chr.18: 3567474-3567474 on Build GRCh38
rs780984705 Chr.18: 3567519-3567519 on Build GRCh38
rs774516068 Chr.18: 3567606-3567606 on Build GRCh38
rs375657622 Chr.18: 3567468-3567468 on Build GRCh38
rs370998045 Chr.18: 3567614-3567614 on Build GRCh38
rs768880582 Chr.18: 3567597-3567597 on Build GRCh38
rs751508081 Chr.18: 3567516-3567516 on Build GRCh38
rs1046077831 Chr.18: 3567539-3567539 on Build GRCh38
rs762416917 Chr.18: 3567607-3567607 on Build GRCh38
rs915583164 Chr.18: 3567484-3567484 on Build GRCh38

View Full Product Details