Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27504925-27505198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX2
Entrez Gene ID
2053
Gene Name
epoxide hydrolase 2
Gene Aliases
CEH, SEH
UniGene
Hs.212088
Gene Chromosome Location
Chr.8: 27491002-27597559 on Build GRCh38
Transcripts
NM_001256482.1 NM_001256483.1 NM_001256484.1 NM_001979.5 XM_017013199.1 XM_017013200.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655312 c.426G>T p.E142D Chr.8: 27505035-27505035 on Build GRCh38
1251386 c.500A>C p.K167T Chr.8: 27505109-27505109 on Build GRCh38
3412962 c.459G>A p.S153S Chr.8: 27505068-27505068 on Build GRCh38
1643585 c.347-9T>C p.? Chr.8: 27504947-27504947 on Build GRCh38
5063083 c.428T>C p.L143P Chr.8: 27505037-27505037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375747773 Chr.8: 27505132-27505132 on Build GRCh38
rs76933761 Chr.8: 27505098-27505098 on Build GRCh38
rs144205298 Chr.8: 27505078-27505078 on Build GRCh38
rs773687108 Chr.8: 27505001-27505001 on Build GRCh38
rs112649081 Chr.8: 27505151-27505151 on Build GRCh38
rs376426011 Chr.8: 27505004-27505004 on Build GRCh38
rs190054509 Chr.8: 27505096-27505096 on Build GRCh38
rs68120452 Chr.8: 27505171-27505171 on Build GRCh38
rs779867120 Chr.8: 27505121-27505121 on Build GRCh38
rs750136903 Chr.8: 27505110-27505110 on Build GRCh38
rs758190602 Chr.8: 27505116-27505116 on Build GRCh38
rs146337543 Chr.8: 27505068-27505068 on Build GRCh38
rs907199664 Chr.8: 27504970-27504970 on Build GRCh38
rs772718243 Chr.8: 27505161-27505161 on Build GRCh38
rs754970262 Chr.8: 27505129-27505129 on Build GRCh38
rs142716458 Chr.8: 27505137-27505137 on Build GRCh38
rs757093870 Chr.8: 27504955-27504955 on Build GRCh38
rs200543697 Chr.8: 27505075-27505075 on Build GRCh38
rs771292961 Chr.8: 27505150-27505150 on Build GRCh38
rs1044409070 Chr.8: 27504961-27504961 on Build GRCh38
rs139657060 Chr.8: 27504998-27504998 on Build GRCh38
rs751495938 Chr.8: 27505128-27505128 on Build GRCh38
rs747665800 Chr.8: 27505135-27505135 on Build GRCh38
rs752932715 Chr.8: 27505029-27505029 on Build GRCh38
rs767564027 Chr.8: 27505058-27505058 on Build GRCh38
rs775096417 Chr.8: 27505154-27505154 on Build GRCh38
rs372428836 Chr.8: 27505061-27505061 on Build GRCh38
rs774803950 Chr.8: 27504991-27504991 on Build GRCh38
rs778807078 Chr.8: 27504978-27504978 on Build GRCh38
rs57699806 Chr.8: 27505070-27505070 on Build GRCh38
rs768249126 Chr.8: 27504994-27504994 on Build GRCh38
rs369553898 Chr.8: 27505030-27505030 on Build GRCh38
rs766917110 Chr.8: 27505005-27505005 on Build GRCh38
rs774974108 Chr.8: 27505090-27505090 on Build GRCh38
rs373754534 Chr.8: 27504969-27504969 on Build GRCh38
rs868305103 Chr.8: 27505126-27505126 on Build GRCh38
rs775644820 Chr.8: 27505162-27505162 on Build GRCh38
rs772104420 Chr.8: 27504986-27504986 on Build GRCh38
rs146049019 Chr.8: 27505106-27505106 on Build GRCh38
rs759707628 Chr.8: 27505015-27505015 on Build GRCh38
rs140021176 Chr.8: 27505109-27505109 on Build GRCh38
rs747000602 Chr.8: 27505067-27505067 on Build GRCh38
rs751645394 Chr.8: 27505009-27505009 on Build GRCh38
rs149400070 Chr.8: 27505060-27505060 on Build GRCh38
rs745786326 Chr.8: 27504984-27504984 on Build GRCh38
rs543103524 Chr.8: 27505146-27505146 on Build GRCh38
rs746553840 Chr.8: 27504993-27504993 on Build GRCh38
rs146853169 Chr.8: 27504997-27504997 on Build GRCh38
rs750246458 Chr.8: 27505043-27505043 on Build GRCh38
rs760157983 Chr.8: 27505094-27505094 on Build GRCh38
rs769295140 Chr.8: 27505136-27505136 on Build GRCh38
rs771290052 Chr.8: 27505083-27505083 on Build GRCh38
rs557211599 Chr.8: 27505160-27505160 on Build GRCh38

View Full Product Details